Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MEFNPSDHPR ASTIFLSKSQ TDVREKRKSL FINHHPPGQI ARKYSSCSTI FLDDSTVSQP NLKYTIKCVA LAIYYHIKNR DPDGRMLLDI FDENLHPLSK SEVPPDYDKH NPEQKQIYRF VRTLFSAAQL TAECAIVTLV YLERLLTYAE IDICPANWKR IVLGAILLAS KVWDDQAVWN VDYCQILKDI TVEDMNELER QFLELLQFNI NVPSSVYAKY YFDLRSLAEA NNLSFPLEPL SRERAHKLEA ISRLCEDKYK DLRRSARKRS ASADNLTLPR WSPAIIS
Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyc-Y; Cyclin box protein 1; Cyclin fold protein 1; cyclin-box carrying protein 1; cyclin-X; Cyclin-Y
Gene Aliases: 1700025H17Rik; 3110050L10Rik; 4631402G10Rik; 5730405I09Rik; C10orf9; CBCP1; CCNX; CCNY; CFP1; RGD1565969
UniProt ID: (Human) Q8ND76, (Mouse) Q8BGU5
Entrez Gene ID: (Human) 219771, (Mouse) 67974, (Rat) 361261
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support