Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyc-Y; Cyclin box protein 1; Cyclin fold protein 1; cyclin-box carrying protein 1; cyclin-X; Cyclin-Y
Gene Aliases: C10orf9; CBCP1; CCNX; CCNY; CFP1
UniProt ID: (Human) Q8ND76
Entrez Gene ID: (Human) 219771
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support