Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total CCRL1.
Receptors for the C-C chemokine family include CCR-1, CCR-2A, CCR-3, CCR-4, CCR-5 and the Duffy blood group antigen. The C-C receptors are important in the function of T cell chemotaxis and migration of phagocytic cells to sites of inflammation. CCR11 in particular is expressed in the heart, lung, and small intestine. It binds members of the monocyte chemoattractant family including MCP-1, MCP-2, and MCP-4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Atypical chemokine receptor 4; C-C chemokine receptor type 11; C-C CKR-11; CC chemokine receptor-like 1; CC-CKR-11; CCR-11; CCRL1; CCX CKR; chemocentryx chemokine receptor; chemokine (C-C motif) receptor-like 1; chemokine (C-C) receptor-like 1; chemokine receptor CCR11; chemokine, cc motif, receptor-like protein 1; orphan seven-transmembrane receptor, chemokine related
Gene Aliases: A630091E18Rik; ACKR4; CC-CKR-11; CCBP2; CCR-11; CCR10; CCR11; CCRL1; CCX CKR; CCX-CKR; CCX-CKR1; CKR-11; Cmkbrl1; PPR1; VSHK1
UniProt ID: (Human) Q9NPB9, (Mouse) Q924I3
Entrez Gene ID: (Human) 51554, (Rat) 685243, (Mouse) 252837
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support