Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCR4 is a chemokine receptor and is preferentially expressed on type 2 helper T (Th2-type) cells. In contrast to other chemokine receptors, the expression of CCR4 and CCR8 on Th2 cells is transiently increased following TCR and CD28 engagement. Moreover, activated Th1 cells up-regulate CCR4 expression and functional responsiveness to thymus- and activation-regulated chemokine. Chemokines are a group of small (approximately 8 to 14 kD), mostly basic, structurally related molecules that regulate cell trafficking of various types of leukocytes through interactions with a subset of 7-transmembrane, G protein-coupled receptors. Chemokines also play fundamental roles in the development, homeostasis, and function of the immune system, and they have effects on cells of the central nervous system as well as on endothelial cells involved in angiogenesis or angiostasis. Chemokines are divided into 2 major subfamilies, CXC and CC, based on the arrangement of the first 2 of the 4 conserved cysteine residues; the 2 cysteines are separated by a single amino acid in CXC chemokines and are adjacent in CC chemokines.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C chemokine receptor 4; C-C chemokine receptor type 4; C-C CKR-4; CCR-4; CD194; chemokine (C-C motif) receptor 4; chemokine (C-C) receptor 4; fusin; K5-5; leukocyte-expressed seven-transmembrane-domain receptor; MGC88293
Gene Aliases: C-C CKR-4; CC-CKR-4; CCR4; CD194; CHEMR1; ChemR13; CKR4; CMKBR4; HGCN:14099; K5-5; LESTR; Sdf1r
UniProt ID: (Human) P51679, (Mouse) P51680
Entrez Gene ID: (Human) 1233, (Rat) 171054, (Mouse) 12773
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support