Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MADDYGSEST SSMEDYVNFN FTDFYCEKNN VRQFASHFL
Antigen sequence identity: Mouse 64%; Rat 67%
The protein encoded by this gene is a member of the beta chemokine receptor family. It is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptors are key regulators of the thymocytes migration and maturation in normal and inflammation conditions. The specific ligand of this receptor is CCL25. It has been found that this gene is differentially expressed by T lymphocytes of small intestine and colon, suggested a role in the thymocytes recruitment and development that may permit functional specialization of immune responses in different segment of the gastrointestinal tract. This gene is mapped to the chemokine receptor gene cluster region. Two alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C chemokine receptor type 9; C-C CKR-9; CDw199; CDw199 antigen; chemokine (C-C motif) receptor 9; G protein-coupled receptor 28; G-protein coupled receptor 28; GPR-9-6
Gene Aliases: CC-CKR-9; CCR9; CDw199; GPR-9-6; GPR28
UniProt ID: (Human) P51686
Entrez Gene ID: (Human) 10803
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support