Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Specificity: 9B1 antibody recognizes CCR9 at the N-terminus. CCR9 is a member of the G protein coupled receptor family and is involved in T cell development in the thymus and in gut-associated immune response. Mouse CCR9 is highly expressed on different stages of thymocytes and upregulated on CD4+CD8+ cells.
The protein encoded by this gene is a member of the beta chemokine receptor family. It is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptors are key regulators of the thymocytes migration and maturation in normal and inflammation conditions. The specific ligand of this receptor is CCL25. It has been found that this gene is differentially expressed by T lymphocytes of small intestine and colon, suggested a role in the thymocytes recruitment and development that may permit functional specialization of immune responses in different segment of the gastrointestinal tract. This gene is mapped to the chemokine receptor gene cluster region. Two alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-C chemokine receptor type 9; C-C CKR-9; CC-CKR-9; CCR-9; CDw199; CDw199 antigen; Chemokine C-C receptor 10
Gene Aliases: A130091K22Rik; Ccr9; Cmkbr10; GPR-9-6
UniProt ID: (Mouse) Q9WUT7
Entrez Gene ID: (Mouse) 12769
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support