Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LICNVSLEYE KTEVNSGFFY KTAEEKEKLV KAERKFIEDR VQKIIDLKDK VCAQSNKGFV VINQKGIDPF SLDSLAKHGI VALRRAKRRN MERLSLACGG MAVNSFEDLT VDCLGHAGLV YEYTLGEEKF TFIEECVNPC SVTLLVKGPN KHTLTQVKDA IRDGLRAIKN AIEDGCMVPG AGAIEVAMAE ALVTYKNSIK GRARLGVQAF ADALLIIPKV LAQNAGYDPQ ETLVKVQAEH VESKQLVGVD LNTGEPMVAA DAGVWDNYCV KKQLLHSCTV IATNILLVDE IMRAGMSSLK
Molecular chaperone; assists the folding of proteins upon ATP hydrolysis. Known to play a role, in vitro, in the folding of actin and tubulin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCT-zeta-2; CCT-zeta-like; Cctz-2; Chaperonin containing T-complex polypeptide 1 subunit 6B; chaperonin containing TCP1, subunit 6B (zeta 2); chaperonin subunit 6b (zeta); chaperonin-containing T-complex polypeptide 1, subunit 6B; T-complex protein 1 subunit zeta-2; TCP-1-zeta-2; TCP-1-zeta-like; Testis-specific protein TSA303; Testis-specific Tcp20
Gene Aliases: CCT-zeta-2; CCT6B; CCTZ-2; Cctz2; CCTzeta-2; TCP-1-zeta-2; TSA303
UniProt ID: (Human) Q92526, (Mouse) Q61390
Entrez Gene ID: (Human) 10693, (Rat) 363658, (Mouse) 12467
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support