Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: RTIPGKQQLL IGAYAKALEI IPRQLCDNAG FDATNILNKL RARHAQGGTW YGVDINNEDI ADNFEAFVWE PAMVRINALT AASEAACLIV SVDETIKNPR STVD
The TCP1 eta gene encodes a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 5 and 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCT-eta; Chaperonin containing T-complex polypeptide 1 subunit 7; chaperonin containing t-complex polypeptide 1, eta subunit; chaperonin containing t-complex subunit eta; chaperonin subunit 7 (eta); HIV-1 Nef interacting protein; HIV-1 Nef-interacting protein; MGC110985; T-complex protein 1 subunit eta; TCP-1-eta
Gene Aliases: AA408524; AL022769; CCT7; CCTETA; CCTH; Cctz; NIP7-1; TCP1ETA
UniProt ID: (Human) Q99832, (Mouse) P80313
Entrez Gene ID: (Human) 10574, (Mouse) 12468, (Rat) 297406
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support