Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Chicken, Bovine and Orangutan
TCP-1 theta is theta subunit of the CCT (chaperonin containing TCP1 complex) chaperonin molecule which is abundant in the eukaryotic cytosol and may be involved in the transport and assembly of the newly synthesized proteins. Molecular chaperones assist in the protein folding upon ATP hydrolysis and TCP-1 theta may play a role in the assembly of the BBsome, a complex that is involved in ciliogenesis regulating transport vesicles to the cilia. Further, TCP-1 theta is also involved in the folding of actin and tubulin. The intact CCT complex is composed of eight polypeptides in a double ring structure. CCT is important within cells in the folding of the VHL tumor suppressor protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCT-theta; Chaperonin containing T-complex polypeptide 1 subunit 8; chaperonin containing TCP1, subunit 8 (theta); chaperonin subunit 8 (theta); Renal carcinoma antigen NY-REN-15; T-complex protein 1 subunit theta; TCP-1-theta
Gene Aliases: AI132397; C21orf112; CCT8; CCTQ; D21S246; KIAA0002; PRED71; Tcpq
UniProt ID: (Human) P50990, (Mouse) P42932
Entrez Gene ID: (Human) 10694, (Mouse) 12469
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support