Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQGPPLLTAA HLLCVCTAAL AVAPGPRFLV TAPGIIRPGG NVTIGVELLE HCPSQVTVKA ELLKTASNLT VSVLEAEGVF EKGSFKTLTL PSLPLNSADE IYELRVTGRT QDEILFSNST RLSFETKRIS VFIQTDKALY KPKQEVKFRI VTLFSDFKPY KTSLNILIKD PKSNLIQQWL SQQSDLGVIS KTFQLSSHPI LGDWSIQVQV NDQTYYQSFQ VSEYVLPKFE VTLQTPLYCS MNSKHLNGTI TAKYTYGKPV KGDVTLTFLP LSFWGKKKNI TKTFKINGSA NFSFNDEEMK NVMDSSNGLS EYLDLSSPGP VEILTTVTES VTGISRNVST NVFFKQHDYI IEFFDYTTVL KPSLNFTATV KVTRADGNQL TLEERRNNVV ITVTQRNYTE YWSGSNSGNQ KMEAVQKINY TVPQSGTFKI EFPILEDSSE LQLKAYFLGS KSSMAVHSLF KSPSKTYIQL KTRDENIKVG SPFELVVSGN KRLKELSYMV VSRGQLVAVG KQNSTMFSLT PENSWTPKAC VIVYYIEDDG EIISDVLKIP VQLVFKNKIK LYWSKVKAEP SEKVSLRISV TQPDSIVGIV AVDKSVNLMN ASNDITMENV VHELELYNTG YYLGMFMNSF AVFQECGLWV LTDANLTKDY IDGVYDNAEY AERFMEENEG HIVDIHDFSL GSSPHVRKHF PETWIWLDTN MGYRIYQEFE VTVPDSITSW VATGFVISED LGLGLTTTPV ELQAFQPFFI FLNLPYSVIR GEEFALEITI FNYLKDATEV KVIIEKSDKF DILMTSNEIN ATGHQQTLLV PSEDGATVLF PIRPTHLGEI PITVTALSPT ASDAVTQMIL VKAEGIEKSY SQSILLDLTD NRLQSTLKTL SFSFPPNTVT GSERVQITAI GDVLGPSING LASLIRMPYG CGEQNMINFA PNIYILDYLT KKKQLTDNLK EKALSFMRQG YQRELLYQRE DGSFSAFGNY DPSGSTWLSA FVLRCFLEAD PYIDIDQNVL HRTYTWLKGH QKSNGEFWDP GRVIHSELQG GNKSPVTLTA YIVTSLLGYR KYQPNIDVQE SIHFLESEFS RGISDNYTLA LITYALSSVG SPKAKEALNM LTWRAEQEGG MQFWVSSESK LSDSWQPRSL DIEVAAYALL SHFLQFQTSE GIPIMRWLSR QRNSLGGFAS TQDTTVALKA LSEFAALMNT ERTNIQVTVT GPSSPSPVKF LIDTHNRLLL QTAELAVVQP TAVNISANGF GFAICQLNVV YNVKASGSSR RRRSIQNQEA FDLDVAVKEN KDDLNHVDLN VCTSFSGPGR SGMALMEVNL LSGFMVPSEA ISLSETVKKV EYDHGKLNLY LDSVNETQFC VNIPAVRNFK VSNTQDASVS IVDYYEPRRQ AVRSYNSEVK LSSCDLCSDV QGCRPCEDGA SGSHHHSSVI FIFCFKLLYF MELWL
CD109 is a member of the alpha2-macroglobulin/complement superfamily. The encoded GPI-linked glycoprotein is found on the cell surface of platelets, activated T-cells, and endothelial cells. The protein binds to and negatively regulates signaling of transforming growth factor beta (TGF-beta).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 150 kDa TGF-beta-1-binding protein; activated T-cell marker CD109; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; CD109; CD109 antigen; Gov platelet alloantigens; p180; Platelet-specific Gov antigen; r150; RP11-525G3.1
Gene Aliases: CD109; CPAMD7; p180; r150
UniProt ID: (Human) Q6YHK3
Entrez Gene ID: (Human) 135228
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support