Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Leinco Technologies
Specificity: Clone GIR-208 recognizes an epitope on human IFN-gamma R-alpha.
Endotoxin Level: ≤ 1.0 EU/mg as determined by the LAL method. Purity: >95% by SDS Page; ≥95% monomer by analytical SEC.
Product Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using only In vitro protein free cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Storage and Handling: Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80 C. Avoid Repeated Freeze Thaw Cycles.
This antibody has been tested in the following applications: Functional Assay, Flow Cytometry.
This gene (IFNGR1) encodes the ligand-binding chain (alpha) of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. A genetic variation in IFNGR1 is associated with susceptibility to Helicobacter pylori infection. In addition, defects in IFNGR1 are a cause of mendelian susceptibility to mycobacterial disease, also known as familial disseminated atypical mycobacterial infection.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CDw119; IFN-gamma receptor 1; IFN-gamma-R-alpha; IFN-gamma-R1; immune interferon receptor 1; Interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor alpha chain
Gene Aliases: CD119; IFNGR; IFNGR1; IMD27A; IMD27B
UniProt ID: (Human) P15260
Entrez Gene ID: (Human) 3459
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support