Product References
Astrocytic interleukin-3 programs microglia and limits Alzheimer's disease.
Nature
McAlpine CS,Park J,Griciuc A,Kim E,Choi SH,Iwamoto Y,Kiss MG,Christie KA,Vinegoni C,Poller WC,Mindur JE,Chan CT,He S,Janssen H,Wong LP,Downey J,Singh S,Anzai A,Kahles F,Jorfi M,Feruglio PF,Sadreyev RI,Weissleder R,Kleinstiver BP,Nahrendorf M,Tanzi RE,Swirski FK
14-1239-82 was used in Immunohistochemistry, Flow Cytometry to identify IL-3 as a key mediator of astrocyte-microglia cross-talk and a node for therapeutic intervention in Alzheimer's disease.
Thu Jul 01 00:00:00 EDT 2021
Astrocytic interleukin-3 programs microglia and limits Alzheimer's disease.
Nature
McAlpine CS,Park J,Griciuc A,Kim E,Choi SH,Iwamoto Y,Kiss MG,Christie KA,Vinegoni C,Poller WC,Mindur JE,Chan CT,He S,Janssen H,Wong LP,Downey J,Singh S,Anzai A,Kahles F,Jorfi M,Feruglio PF,Sadreyev RI,Weissleder R,Kleinstiver BP,Nahrendorf M,Tanzi RE,Swirski FK
14-1239-82 was used in Immunohistochemistry, Flow Cytometry to identify IL-3 as a key mediator of astrocyte-microglia cross-talk and a node for therapeutic intervention in Alzheimer's disease.
Thu Jul 01 00:00:00 EDT 2021
Astrocytic interleukin-3 programs microglia and limits Alzheimer's disease.
Nature
McAlpine CS,Park J,Griciuc A,Kim E,Choi SH,Iwamoto Y,Kiss MG,Christie KA,Vinegoni C,Poller WC,Mindur JE,Chan CT,He S,Janssen H,Wong LP,Downey J,Singh S,Anzai A,Kahles F,Jorfi M,Feruglio PF,Sadreyev RI,Weissleder R,Kleinstiver BP,Nahrendorf M,Tanzi RE,Swirski FK
14-1239-82 was used in Immunohistochemistry, Flow Cytometry to identify IL-3 as a key mediator of astrocyte-microglia cross-talk and a node for therapeutic intervention in Alzheimer's disease.
Thu Jul 01 00:00:00 EDT 2021
Astrocytic interleukin-3 programs microglia and limits Alzheimer's disease.
Nature
McAlpine CS,Park J,Griciuc A,Kim E,Choi SH,Iwamoto Y,Kiss MG,Christie KA,Vinegoni C,Poller WC,Mindur JE,Chan CT,He S,Janssen H,Wong LP,Downey J,Singh S,Anzai A,Kahles F,Jorfi M,Feruglio PF,Sadreyev RI,Weissleder R,Kleinstiver BP,Nahrendorf M,Tanzi RE,Swirski FK
14-1239-82 was used in Immunohistochemistry, Flow Cytometry to identify IL-3 as a key mediator of astrocyte-microglia cross-talk and a node for therapeutic intervention in Alzheimer's disease.
Thu Jul 01 00:00:00 EDT 2021
Unsupervised Analysis of Flow Cytometry Data in a Clinical Setting Captures Cell Diversity and Allows Population Discovery.
Frontiers in immunology
Baumgaertner P,Sankar M,Herrera F,Benedetti F,Barras D,Thierry AC,Dangaj D,Kandalaft LE,Coukos G,Xenarios I,Guex N,Harari A
Published figure using CD123 monoclonal antibody (Product # 14-1239-82) in Flow Cytometry
Tue Sep 21 00:00:00 EDT 2021
Single-Cell Analyses Reveal Megakaryocyte-Biased Hematopoiesis in Myelofibrosis and Identify Mutant Clone-Specific Targets.
Molecular cell
Psaila B,Wang G,Rodriguez-Meira A,Li R,Heuston EF,Murphy L,Yee D,Hitchcock IS,Sousos N,O'Sullivan J,Anderson S,Senis YA,Weinberg OK,Calicchio ML,Iskander D,Royston D,Milojkovic D,Roberts I,Bodine DM,Thongjuea S,Mead AJ
Published figure using CD123 monoclonal antibody (Product # 14-1239-82) in Flow Cytometry
Thu May 07 00:00:00 EDT 2020
Systems Biology Methods Applied to Blood and Tissue for a Comprehensive Analysis of Immune Response to Hepatitis B Vaccine in Adults.
Frontiers in immunology
Ben-Othman R,Cai B,Liu AC,Varankovich N,He D,Blimkie TM,Lee AH,Gill EE,Novotny M,Aevermann B,Drissler S,Shannon CP,McCann S,Marty K,Bjornson G,Edgar RD,Lin DTS,Gladish N,Maclsaac J,Amenyogbe N,Chan Q,Llibre A,Collin J,Landais E,Le K,Reiss SM,Koff WC,Havenar-Daughton C,Heran M,Sangha B,Walt D,Krajden M,Crotty S,Sok D,Briney B,Burton DR,Duffy D,Foster LJ,Mohn WW,Kobor MS,Tebbutt SJ,Brinkman RR,Scheuermann RH,Hancock REW,Kollmann TR,Sadarangani M
Published figure using CD123 monoclonal antibody (Product # 14-1239-82) in Flow Cytometry
Mon Jun 21 00:00:00 EDT 2021
Human pDCs Are Superior to cDC2s in Attracting Cytolytic Lymphocytes in Melanoma Patients Receiving DC Vaccination.
Cell reports
van Beek JJP,Flórez-Grau G,Gorris MAJ,Mathan TSM,Schreibelt G,Bol KF,Textor J,de Vries IJM
14-1239 was used in Flow cytometry/Cell sorting to show that pDCs and cDC2s display a striking difference in chemokine secretion, which correlates with the recruitment of distinct types of immune effector cells.
Tue Jan 28 00:00:00 EST 2020
Quantifying in situ adaptive immune cell cognate interactions in humans.
Nature immunology
Liarski VM,Sibley A,van Panhuys N,Ai J,Chang A,Kennedy D,Merolle M,Germain RN,Giger ML,Clark MR
14-1239 was used in Immunohistochemistry to study the specificity and characteristics of liver sinusoidal endothelial cells compared to lung and heart endothelial cells.
Mon Apr 01 00:00:00 EDT 2019
Neutrophils Inhibit Synthesis of Mineralized Extracellular Matrix by Human Bone Marrow-Derived Stromal Cells In Vitro.
Frontiers in immunology
Bastian OW,Croes M,Alblas J,Koenderman L,Leenen LPH,Blokhuis TJ
Published figure using CD123 monoclonal antibody (Product # 14-1239-82) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
High Expression of Galectin-3 in Patients with IgG4-Related Disease: A Proteomic Approach.
Pathology research international
Salah A,Yoshifuji H,Ito S,Kitagori K,Kiso K,Yamada N,Nakajima T,Haga H,Tsuruyama T,Miyagawa-Hayashino A
14-1239 was used in Immunohistochemistry to study protein expression profiles and identify a potential biomarker for IgG4-related disease.
Wed Sep 30 00:00:00 EDT 2020
Self-reactive IgE exacerbates interferon responses associated with autoimmunity.
Nature immunology
Henault J,Riggs JM,Karnell JL,Liarski VM,Li J,Shirinian L,Xu L,Casey KA,Smith MA,Khatry DB,Izhak L,Clarke L,Herbst R,Ettinger R,Petri M,Clark MR,Mustelin T,Kolbeck R,Sanjuan MA
14-1239 was used in Immunofluorescence on frozen tissues to investigate the role of IgE in systemic lupus erythematosus.
Mon Feb 01 00:00:00 EST 2016
Humanized Mice Reveal Differential Immunogenicity of Cells Derived from Autologous Induced Pluripotent Stem Cells.
Cell stem cell
Zhao T,Zhang ZN,Westenskow PD,Todorova D,Hu Z,Lin T,Rong Z,Kim J,He J,Wang M,Clegg DO,Yang YG,Zhang K,Friedlander M,Xu Y
14-1239 was used in Immunohistochemistry on frozen tissues to demonstrate that most teratomas formed by autologous integration-free hiPSCs exhibit local infiltration of antigen-specific T-cells and associated tissue necrosis.
Thu Sep 03 00:00:00 EDT 2015
Brief Report: IFIH1 Mutation Causes Systemic Lupus Erythematosus With Selective IgA Deficiency.
Arthritis & rheumatology (Hoboken, N.J.)
Van Eyck L,De Somer L,Pombal D,Bornschein S,Frans G,Humblet-Baron S,Moens L,de Zegher F,Bossuyt X,Wouters C,Liston A
14-1239 was used in Flow cytometry/Cell sorting to determine the underlying genetic defect responsible for severe early onset and refractory systemic lupus erythematosus.
Mon Jun 01 00:00:00 EDT 2015
Thymic HIV-2 infection uncovers posttranscriptional control of viral replication in human thymocytes.
Journal of virology
Nunes-Cabaço H,Matoso P,Foxall RB,Tendeiro R,Pires AR,Carvalho T,Pinheiro AI,Soares RS,Sousa AE
14-1239 was used in Flow cytometry/Cell sorting to assess the direct impact of HIV-2 infection on the human thymus.
Sun Feb 01 00:00:00 EST 2015
Comparative transcriptional and functional profiling defines conserved programs of intestinal DC differentiation in humans and mice.
Nature immunology
Watchmaker PB,Lahl K,Lee M,Baumjohann D,Morton J,Kim SJ,Zeng R,Dent A,Ansel KM,Diamond B,Hadeiba H,Butcher EC
14-1239 was used in Flow cytometry/Cell sorting to characterise human gut dendritic cell populations, and define their relationship to previously studied human and mouse dendritic cells.
Wed Jan 01 00:00:00 EST 2014
Malaria-induced NLRP12/NLRP3-dependent caspase-1 activation mediates inflammation and hypersensitivity to bacterial superinfection.
PLoS pathogens
Ataide MA,Andrade WA,Zamboni DS,Wang D,Souza Mdo C,Franklin BS,Elian S,Martins FS,Pereira D,Reed G,Fitzgerald KA,Golenbock DT,Gazzinelli RT
Published figure using CD123 monoclonal antibody (Product # 14-1239-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2014
Lipoxin A4 regulates natural killer cell and type 2 innate lymphoid cell activation in asthma.
Science translational medicine
Barnig C,Cernadas M,Dutile S,Liu X,Perrella MA,Kazani S,Wechsler ME,Israel E,Levy BD
14-1239 was used in Flow cytometry/Cell sorting to identify the potential of innate lymphoid cells to decrease airway inflammation.
Wed Feb 27 00:00:00 EST 2013
R-phycoerythrin-conjugated antibodies are inappropriate for intracellular staining of murine plasma cells.
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Kim MS,Kim TS
Published figure using CD123 monoclonal antibody (Product # 14-1239-82) in Flow Cytometry
Wed May 01 00:00:00 EDT 2013
Frailty in old age is associated with decreased interleukin-12/23 production in response to toll-like receptor ligation.
PloS one
Compté N,Zouaoui Boudjeltia K,Vanhaeverbeek M,De Breucker S,Tassignon J,Trelcat A,Pepersack T,Goriely S
Published figure using CD123 monoclonal antibody (Product # 14-1239-82) in Flow Cytometry
Tue Jan 14 00:00:00 EST 2014
Possible implication of local immune response in Darier's disease: an immunohistochemical characterization of lesional inflammatory infiltrate.
Mediators of inflammation
Miracco C,Pietronudo F,Mourmouras V,Pellegrino M,Onorati M,Mastrogiulio MG,Cantarini L,Luzi P
14-1239 was used in Immunohistochemistry to characterize and quantify the local inflammatory infiltrate in Darier's Disease skin lesions.
Thu Nov 04 00:00:00 EDT 2010
Irreversible loss of pDCs by apoptosis during early HIV infection may be a critical determinant of immune dysfunction.
Viral immunology
Meera S,Madhuri T,Manisha G,Ramesh P
Published figure using CD123 monoclonal antibody (Product # 14-1239-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2010
Lung dendritic cell expression of maturation molecules increases with worsening chronic obstructive pulmonary disease.
American journal of respiratory and critical care medicine
Freeman CM,Martinez FJ,Han MK,Ames TM,Chensue SW,Todt JC,Arenberg DA,Meldrum CA,Getty C,McCloskey L,Curtis JL
14-1239 was used in Flow cytometry/Cell sorting to determine the expression of maturation molecules by individual DC subsets in relationship to COPD stage.
Tue Dec 15 00:00:00 EST 2009
Human spleen contains different subsets of dendritic cells and regulatory T lymphocytes.
Clinical and experimental immunology
Velásquez-Lopera MM,Correa LA,García LF
14-1239-82 was used in Immunohistochemistry to shows the frequency, phenotype and location of DCs and regulatory T cells in deceased organ donors' spleens.
Wed Oct 01 00:00:00 EDT 2008
Epithelial cells trigger frontline immunoglobulin class switching through a pathway regulated by the inhibitor SLPI.
Nature immunology
Xu W,He B,Chiu A,Chadburn A,Shan M,Buldys M,Ding A,Knowles DM,Santini PA,Cerutti A
Thu Mar 01 00:00:00 EST 2007
Local activation of the innate immune system in Buruli ulcer lesions.
The Journal of investigative dermatology
Peduzzi E,Groeper C,Schütte D,Zajac P,Rondini S,Mensah-Quainoo E,Spagnoli GC,Pluschke G,Daubenberger CA
Thu Mar 01 00:00:00 EST 2007
CD56-positive haematological neoplasms of the skin: a multicentre study of the Cutaneous Lymphoma Project Group of the European Organisation for Research and Treatment of Cancer.
Journal of clinical pathology
Assaf C,Gellrich S,Whittaker S,Robson A,Cerroni L,Massone C,Kerl H,Rose C,Chott A,Chimenti S,Hallermann C,Petrella T,Wechsler J,Bagot M,Hummel M,Bullani-Kerl K,Bekkenk MW,Kempf W,Meijer CJ,Willemze R,Sterry W
Sat Sep 01 00:00:00 EDT 2007
CD56-positive haematological neoplasms of the skin: a multicentre study of the Cutaneous Lymphoma Project Group of the European Organisation for Research and Treatment of Cancer.
Journal of clinical pathology
Assaf C,Gellrich S,Whittaker S,Robson A,Cerroni L,Massone C,Kerl H,Rose C,Chott A,Chimenti S,Hallermann C,Petrella T,Wechsler J,Bagot M,Hummel M,Bullani-Kerl K,Bekkenk MW,Kempf W,Meijer CJ,Willemze R,Sterry W
Sat Sep 01 00:00:00 EDT 2007
Expression of the plasmacytoid dendritic cell marker BDCA-2 supports a spectrum of maturation among CD4+ CD56+ hematodermic neoplasms.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Jaye DL,Geigerman CM,Herling M,Eastburn K,Waller EK,Jones D
Fri Dec 01 00:00:00 EST 2006