Product References
LncRNA BC200/miR-150-5p/MYB positive feedback loop promotes the malignant proliferation of myelodysplastic syndrome.
Cell death & disease
Liu Z,Wang P,Yuan S,Wang Y,Cao P,Wen F,Li H,Zhu L,Liang L,Wang Z,Hu B,Zheng F,Liu J,Xiao X,Zhang J
25-1239-42 was used in Flow Cytometry to investigate the molecular mechanisms of the specific roles of long noncoding RNAs (lncRNAs) in Myelodysplastic syndrome.
Tue Feb 08 00:00:00 EST 2022
Unsupervised Analysis of Flow Cytometry Data in a Clinical Setting Captures Cell Diversity and Allows Population Discovery.
Frontiers in immunology
Baumgaertner P,Sankar M,Herrera F,Benedetti F,Barras D,Thierry AC,Dangaj D,Kandalaft LE,Coukos G,Xenarios I,Guex N,Harari A
Published figure using CD123 monoclonal antibody (Product # 25-1239-42) in Flow Cytometry
Tue Sep 21 00:00:00 EDT 2021
Transitions in lineage specification and gene regulatory networks in hematopoietic stem/progenitor cells over human development.
Cell reports
Roy A,Wang G,Iskander D,O'Byrne S,Elliott N,O'Sullivan J,Buck G,Heuston EF,Wen WX,Meira AR,Hua P,Karadimitris A,Mead AJ,Bodine DM,Roberts I,Psaila B,Thongjuea S
25-1239-42 was used in Flow cytometry/Cell sorting to demonstrate the utility of this dataset for understanding aberrant hematopoiesis through comparison to two cancers that present at distinct time points in postnatal life-juvenile myelomonocytic leukemia, a childhood cancer, and myelofibrosis, which classically presents in older adults.
Tue Sep 14 00:00:00 EDT 2021
An IDH1-vitamin C crosstalk drives human erythroid development by inhibiting pro-oxidant mitochondrial metabolism.
Cell reports
Gonzalez-Menendez P,Romano M,Yan H,Deshmukh R,Papoin J,Oburoglu L,Daumur M,Dumé AS,Phadke I,Mongellaz C,Qu X,Bories PN,Fontenay M,An X,Dardalhon V,Sitbon M,Zimmermann VS,Gallagher PG,Tardito S,Blanc L,Mohandas N,Taylor N,Kinet S
25-1239-42 was used in Flow cytometry/Cell sorting to show that an IDH1-vitamin C crosstalk controls terminal steps of human erythroid differentiation.
Tue Feb 02 00:00:00 EST 2021
Single-Cell Analyses Reveal Megakaryocyte-Biased Hematopoiesis in Myelofibrosis and Identify Mutant Clone-Specific Targets.
Molecular cell
Psaila B,Wang G,Rodriguez-Meira A,Li R,Heuston EF,Murphy L,Yee D,Hitchcock IS,Sousos N,O'Sullivan J,Anderson S,Senis YA,Weinberg OK,Calicchio ML,Iskander D,Royston D,Milojkovic D,Roberts I,Bodine DM,Thongjuea S,Mead AJ
Published figure using CD123 monoclonal antibody (Product # 25-1239-42) in Flow Cytometry
Thu May 07 00:00:00 EDT 2020
Single-Cell Analyses Reveal Megakaryocyte-Biased Hematopoiesis in Myelofibrosis and Identify Mutant Clone-Specific Targets.
Molecular cell
Psaila B,Wang G,Rodriguez-Meira A,Li R,Heuston EF,Murphy L,Yee D,Hitchcock IS,Sousos N,O'Sullivan J,Anderson S,Senis YA,Weinberg OK,Calicchio ML,Iskander D,Royston D,Milojkovic D,Roberts I,Bodine DM,Thongjuea S,Mead AJ
Published figure using CD123 monoclonal antibody (Product # 25-1239-42) in Flow Cytometry
Thu May 07 00:00:00 EDT 2020
Systems Biology Methods Applied to Blood and Tissue for a Comprehensive Analysis of Immune Response to Hepatitis B Vaccine in Adults.
Frontiers in immunology
Ben-Othman R,Cai B,Liu AC,Varankovich N,He D,Blimkie TM,Lee AH,Gill EE,Novotny M,Aevermann B,Drissler S,Shannon CP,McCann S,Marty K,Bjornson G,Edgar RD,Lin DTS,Gladish N,Maclsaac J,Amenyogbe N,Chan Q,Llibre A,Collin J,Landais E,Le K,Reiss SM,Koff WC,Havenar-Daughton C,Heran M,Sangha B,Walt D,Krajden M,Crotty S,Sok D,Briney B,Burton DR,Duffy D,Foster LJ,Mohn WW,Kobor MS,Tebbutt SJ,Brinkman RR,Scheuermann RH,Hancock REW,Kollmann TR,Sadarangani M
25-1239 was used in Flow cytometry/Cell sorting to escribe a detailed approach that was applied in a study of 15 healthy adults, who were immunized with hepatitis B vaccine.
Mon Jun 21 00:00:00 EDT 2021
Systems Biology Methods Applied to Blood and Tissue for a Comprehensive Analysis of Immune Response to Hepatitis B Vaccine in Adults.
Frontiers in immunology
Ben-Othman R,Cai B,Liu AC,Varankovich N,He D,Blimkie TM,Lee AH,Gill EE,Novotny M,Aevermann B,Drissler S,Shannon CP,McCann S,Marty K,Bjornson G,Edgar RD,Lin DTS,Gladish N,Maclsaac J,Amenyogbe N,Chan Q,Llibre A,Collin J,Landais E,Le K,Reiss SM,Koff WC,Havenar-Daughton C,Heran M,Sangha B,Walt D,Krajden M,Crotty S,Sok D,Briney B,Burton DR,Duffy D,Foster LJ,Mohn WW,Kobor MS,Tebbutt SJ,Brinkman RR,Scheuermann RH,Hancock REW,Kollmann TR,Sadarangani M
25-1239 was used in Flow cytometry/Cell sorting to escribe a detailed approach that was applied in a study of 15 healthy adults, who were immunized with hepatitis B vaccine.
Mon Jun 21 00:00:00 EDT 2021
Immune characterization of metastatic colorectal cancer patients post reovirus administration.
BMC cancer
Parakrama R,Fogel E,Chandy C,Augustine T,Coffey M,Tesfa L,Goel S,Maitra R
25-1239 was used in Flow cytometry/Cell sorting to evaluate the nature of immune response by determining the cytokine expression pattern in peripheral circulation along with the distribution of antigen presenting cells (APCs) and activated T lymphocytes.
Thu Jun 18 00:00:00 EDT 2020
Neutrophils Inhibit Synthesis of Mineralized Extracellular Matrix by Human Bone Marrow-Derived Stromal Cells In Vitro.
Frontiers in immunology
Bastian OW,Croes M,Alblas J,Koenderman L,Leenen LPH,Blokhuis TJ
Published figure using CD123 monoclonal antibody (Product # 25-1239-42) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
Genetic Architecture of Adaptive Immune System Identifies Key Immune Regulators.
Cell reports
Lagou V,Garcia-Perez JE,Smets I,Van Horebeek L,Vandebergh M,Chen L,Mallants K,Prezzemolo T,Hilven K,Humblet-Baron S,Moisse M,Van Damme P,Boeckxstaens G,Bowness P,Dubois B,Dooley J,Liston A,Goris A
25-1239-42 was used in Flow Cytometry to identify eight genome-wide significant associations explaining 6%-20% of variance in immune system regulation.
Tue Oct 16 00:00:00 EDT 2018
Heterogeneous leukemia stem cells in myeloid blast phase chronic myeloid leukemia.
Blood advances
Kinstrie R,Karamitros D,Goardon N,Morrison H,Hamblin M,Robinson L,Clark RE,Copland M,Vyas P
25-1239 was used in Flow cytometry/Cell sorting to study the haemopoietic hierarchy of additional mutations in blast phase chronic myeloid leukaemia, and the cellular identity of the resulting leukaemia-propagating stem cell.
Tue Dec 27 00:00:00 EST 2016
Malaria-induced NLRP12/NLRP3-dependent caspase-1 activation mediates inflammation and hypersensitivity to bacterial superinfection.
PLoS pathogens
Ataide MA,Andrade WA,Zamboni DS,Wang D,Souza Mdo C,Franklin BS,Elian S,Martins FS,Pereira D,Reed G,Fitzgerald KA,Golenbock DT,Gazzinelli RT
Published figure using CD123 monoclonal antibody (Product # 25-1239-42) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2014
Target cell availability, rather than breast milk factors, dictates mother-to-infant transmission of SIV in sooty mangabeys and rhesus macaques.
PLoS pathogens
Chahroudi A,Cartwright E,Lee ST,Mavigner M,Carnathan DG,Lawson B,Carnathan PM,Hashempoor T,Murphy MK,Meeker T,Ehnert S,Souder C,Else JG,Cohen J,Collman RG,Vanderford TH,Permar SR,Derdeyn CA,Villinger F,Silvestri G
25-1239 was used in Flow cytometry/Cell sorting to elucidate the factors that prevent the vertical transmission of SIV in sooty mangabeys.
Sat Mar 01 00:00:00 EST 2014
Target cell availability, rather than breast milk factors, dictates mother-to-infant transmission of SIV in sooty mangabeys and rhesus macaques.
PLoS pathogens
Chahroudi A,Cartwright E,Lee ST,Mavigner M,Carnathan DG,Lawson B,Carnathan PM,Hashempoor T,Murphy MK,Meeker T,Ehnert S,Souder C,Else JG,Cohen J,Collman RG,Vanderford TH,Permar SR,Derdeyn CA,Villinger F,Silvestri G
25-1239 was used in Flow cytometry/Cell sorting to elucidate the factors that prevent the vertical transmission of SIV in sooty mangabeys.
Sat Mar 01 00:00:00 EST 2014
Intact type I Interferon production and IRF7 function in sooty mangabeys.
PLoS pathogens
Bosinger SE,Johnson ZP,Folkner KA,Patel N,Hashempour T,Jochems SP,Del Rio Estrada PM,Paiardini M,Lin R,Vanderford TH,Hiscott J,Silvestri G
25-1239 was used in Flow cytometry/Cell sorting to study the levels of Interferon-α and Interferon Regulatory Factor-7 signalling in SIV-infected sooty mangabeys.
Mon Mar 17 00:00:00 EDT 2014
Frailty in old age is associated with decreased interleukin-12/23 production in response to toll-like receptor ligation.
PloS one
Compté N,Zouaoui Boudjeltia K,Vanhaeverbeek M,De Breucker S,Tassignon J,Trelcat A,Pepersack T,Goriely S
25-1239 was used in Flow cytometry/Cell sorting to study the correlation between old age frailty and decreased interleukin-12/23 production due to toll-like receptor ligation.
Tue Jan 14 00:00:00 EST 2014
HIV-1 infection-induced apoptotic microparticles inhibit human DCs via CD44.
The Journal of clinical investigation
Frleta D,Ochoa CE,Kramer HB,Khan SA,Stacey AR,Borrow P,Kessler BM,Haynes BF,Bhardwaj N
25-1239 was used in Flow cytometry/Cell sorting to suggest that targeting CD44-mediated inhibition by apoptotic microparticles could be a novel strategy to potentiate DC activation of HIV-specific immunity.
Sat Dec 01 00:00:00 EST 2012
Interferon regulatory factor modulation underlies the bystander suppression of malaria antigen-driven IL-12 and IFN-γ in filaria-malaria co-infection.
European journal of immunology
Metenou S,Kovacs M,Dembele B,Coulibaly YI,Klion AD,Nutman TB
25-1239 was used in Flow cytometry/Cell sorting to determine mechanisms underlying the suppression of malaria-specific Th1-cell and pro-inflammatory responses in filaria-infected subjects and in vitro.
Thu Mar 01 00:00:00 EST 2012
Generation of bivalent chromatin domains during cell fate decisions.
Epigenetics & chromatin
De Gobbi M,Garrick D,Lynch M,Vernimmen D,Hughes JR,Goardon N,Luc S,Lower KM,Sloane-Stanley JA,Pina C,Soneji S,Renella R,Enver T,Taylor S,Jacobsen SE,Vyas P,Gibbons RJ,Higgs DR
25-1239 was used in Flow cytometry/Cell sorting to investigate the patterns and levels of histone modifications at bivalent genes, showing the generation of bivalent chromatin domains during cell fate decisions.
Mon Jun 06 00:00:00 EDT 2011
Irreversible loss of pDCs by apoptosis during early HIV infection may be a critical determinant of immune dysfunction.
Viral immunology
Meera S,Madhuri T,Manisha G,Ramesh P
Published figure using CD123 monoclonal antibody (Product # 25-1239-42) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2010
Major depletion of plasmacytoid dendritic cells in HIV-2 infection, an attenuated form of HIV disease.
PLoS pathogens
Cavaleiro R,Baptista AP,Soares RS,Tendeiro R,Foxall RB,Gomes P,Victorino RM,Sousa AE
25-1239 was used in Flow cytometry/Cell sorting to understand the contribution of plasmacytoid dendritic cell biology to HIV/AIDS pathogenesis.
Sun Nov 01 00:00:00 EDT 2009
Epithelial cells trigger frontline immunoglobulin class switching through a pathway regulated by the inhibitor SLPI.
Nature immunology
Xu W,He B,Chiu A,Chadburn A,Shan M,Buldys M,Ding A,Knowles DM,Santini PA,Cerutti A
Thu Mar 01 00:00:00 EST 2007
Local activation of the innate immune system in Buruli ulcer lesions.
The Journal of investigative dermatology
Peduzzi E,Groeper C,Schütte D,Zajac P,Rondini S,Mensah-Quainoo E,Spagnoli GC,Pluschke G,Daubenberger CA
Thu Mar 01 00:00:00 EST 2007
CD56-positive haematological neoplasms of the skin: a multicentre study of the Cutaneous Lymphoma Project Group of the European Organisation for Research and Treatment of Cancer.
Journal of clinical pathology
Assaf C,Gellrich S,Whittaker S,Robson A,Cerroni L,Massone C,Kerl H,Rose C,Chott A,Chimenti S,Hallermann C,Petrella T,Wechsler J,Bagot M,Hummel M,Bullani-Kerl K,Bekkenk MW,Kempf W,Meijer CJ,Willemze R,Sterry W
Sat Sep 01 00:00:00 EDT 2007
Expression of the plasmacytoid dendritic cell marker BDCA-2 supports a spectrum of maturation among CD4+ CD56+ hematodermic neoplasms.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Jaye DL,Geigerman CM,Herling M,Eastburn K,Waller EK,Jones D
Fri Dec 01 00:00:00 EST 2006