Product References
CD8α+ dendritic cells potentiate antitumor and immune activities against murine ovarian cancers.
Scientific reports
Lee SW,Lee H,Lee KW,Kim MJ,Kang SW,Lee YJ,Kim H,Kim YM
17-0141-82 was used in Flow cytometry/Cell sorting to report the effectiveness of mouse CD8α+ DCs derived from bone marrow hematopoietic stem cells (BM-HSCs), equivalent to human CD141+ DCs, which have proven to be a highly superior subset.
Tue Jan 03 00:00:00 EST 2023
Macrophage polarization is involved in liver fibrosis induced by β 1-adrenoceptor autoantibody.
Acta biochimica et biophysica Sinica
Wu Y,Fan X,Yu H,Liu J,Duan Y,Zhang S,Yan L,Du Y,Liu H
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Thu Aug 25 00:00:00 EDT 2022
Early activation of the cardiac CX3CL1/CX3CR1 axis delays β-adrenergic-induced heart failure.
Scientific reports
Flamant M,Mougenot N,Balse E,Le Fèvre L,Atassi F,Gautier EL,Le Goff W,Keck M,Nadaud S,Combadière C,Boissonnas A,Pavoine C
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Thu Sep 09 00:00:00 EDT 2021
CD14+CD16- monocytes are the main precursors of osteoclasts in rheumatoid arthritis via expressing Tyro3TK.
Arthritis research & therapy
Xue J,Xu L,Zhu H,Bai M,Li X,Zhao Z,Zhong H,Cheng G,Li X,Hu F,Su Y
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Mon Sep 21 00:00:00 EDT 2020
CNS Immune Profiling in a Dengue Virus-Infected Immunocompetent Outbred ICR Mice Strain.
Frontiers in cellular and infection microbiology
Shen TJ,Chen CL,Jhan MK,Tseng PC,Lin CF
17-0141 was used in Flow cytometry/Cell sorting to indicate a CNS immune profile of DENV infection and hypothetical CNS immunity in response to DENV infection.
Thu Jun 17 00:00:00 EDT 2021
CNS Immune Profiling in a Dengue Virus-Infected Immunocompetent Outbred ICR Mice Strain.
Frontiers in cellular and infection microbiology
Shen TJ,Chen CL,Jhan MK,Tseng PC,Lin CF
17-0141 was used in Flow cytometry/Cell sorting to indicate a CNS immune profile of DENV infection and hypothetical CNS immunity in response to DENV infection.
Thu Jun 17 00:00:00 EDT 2021
A Subset of Skin Macrophages Contributes to the Surveillance and Regeneration of Local Nerves.
Immunity
Kolter J,Feuerstein R,Zeis P,Hagemeyer N,Paterson N,d'Errico P,Baasch S,Amann L,Masuda T,Lösslein A,Gharun K,Meyer-Luehmann M,Waskow C,Franzke CW,Grün D,Lämmermann T,Prinz M,Henneke P
17-0141 was used in Flow cytometry/Cell sorting to delineate the postnatal development of dermal macrophages and their differentiation into subsets by adapting single-cell transcriptomics, fate mapping, and imaging.
Tue Jun 18 00:00:00 EDT 2019
Leucine-rich repeats and calponin homology containing 4 (Lrch4) regulates the innate immune response.
The Journal of biological chemistry
Aloor JJ,Azzam KM,Guardiola JJ,Gowdy KM,Madenspacher JH,Gabor KA,Mueller GA,Lin WC,Lowe JM,Gruzdev A,Henderson MW,Draper DW,Merrick BA,Fessler MB
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Fri Feb 08 00:00:00 EST 2019
Chanzyme TRPM7 Mediates the Ca2+ Influx Essential for Lipopolysaccharide-Induced Toll-Like Receptor 4 Endocytosis and Macrophage Activation.
Immunity
Schappe MS,Szteyn K,Stremska ME,Mendu SK,Downs TK,Seegren PV,Mahoney MA,Dixit S,Krupa JK,Stipes EJ,Rogers JS,Adamson SE,Leitinger N,Desai BN
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Tue Jan 16 00:00:00 EST 2018
TIM‑4 blockade of KCs combined with exogenous TGF‑β injection helps to reverse acute rejection and prolong the survival rate of mice receiving liver allografts.
International journal of molecular medicine
Wu H,Xu X,Li J,Gong J,Li M
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2018
MicroRNA-10a/b are regulators of myeloid differentiation and acute myeloid leukemia.
Oncology letters
Bi L,Sun L,Jin Z,Zhang S,Shen Z
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Butyrate upregulates the TLR4 expression and the phosphorylation of MAPKs and NK-κB in colon cancer cell in vitro.
Oncology letters
Xiao T,Wu S,Yan C,Zhao C,Jin H,Yan N,Xu J,Wu Y,Li C,Shao Q,Xia S
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2018
Measurement of TLR4 and CD14 Receptor Endocytosis Using Flow Cytometry.
Bio-protocol
Schappe MS,Desai BN
17-0141 was used in Flow cytometry/Cell sorting to describe a flow cytometry-based method to study the role of the ion channel TRPM7 in TLR4 endocytosis.
Fri Jul 20 00:00:00 EDT 2018
By Capturing Inflammatory Lipids Released from Dying Cells, the Receptor CD14 Induces Inflammasome-Dependent Phagocyte Hyperactivation.
Immunity
Zanoni I,Tan Y,Di Gioia M,Springstead JR,Kagan JC
17-0141 was used in Flow cytometry/Cell sorting to demonstrate that the CD14 receptor can capture extracellular oxPAPC from dying cells and deliver them into the cell to promote inflammasome-dependent phagocyte hyperactivation.
Tue Oct 17 00:00:00 EDT 2017
Developmental origin and maintenance of distinct testicular macrophage populations.
The Journal of experimental medicine
Mossadegh-Keller N,Gentek R,Gimenez G,Bigot S,Mailfert S,Sieweke MH
17-0141-82 was used in Flow Cytometry to investigate the developmental origins and mode of homeostatic maintenance of two distinct testicular macrophages populations.
Mon Oct 02 00:00:00 EDT 2017
Construction of developmental lineage relationships in the mouse mammary gland by single-cell RNA profiling.
Nature communications
Pal B,Chen Y,Vaillant F,Jamieson P,Gordon L,Rios AC,Wilcox S,Fu N,Liu KH,Jackling FC,Davis MJ,Lindeman GJ,Smyth GK,Visvader JE
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Mon Nov 20 00:00:00 EST 2017
T3 Regulates a Human Macrophage-Derived TSH-β Splice Variant: Implications for Human Bone Biology.
Endocrinology
Baliram R,Latif R,Morshed SA,Zaidi M,Davies TF
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Thu Sep 01 00:00:00 EDT 2016
Murine iPSC-Derived Macrophages as a Tool for Disease Modeling of Hereditary Pulmonary Alveolar Proteinosis due to Csf2rb Deficiency.
Stem cell reports
Mucci A,Kunkiel J,Suzuki T,Brennig S,Glage S,Kühnel MP,Ackermann M,Happle C,Kuhn A,Schambach A,Trapnell BC,Hansen G,Moritz T,Lachmann N
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
Insulin decreases atherosclerosis by inducing endothelin receptor B expression.
JCI insight
Park K,Mima A,Li Q,Rask-Madsen C,He P,Mizutani K,Katagiri S,Maeda Y,Wu IH,Khamaisi M,Preil SR,Maddaloni E,Sørensen D,Rasmussen LM,Huang PL,King GL
17-0141 was used in Flow cytometry/Cell sorting to investigate how accelerated atherosclerosis in diabetes may be reduced by improving insulin signalling.
Thu May 05 00:00:00 EDT 2016
Mechanisms of Toll-like Receptor 4 Endocytosis Reveal a Common Immune-Evasion Strategy Used by Pathogenic and Commensal Bacteria.
Immunity
Tan Y,Zanoni I,Cullen TW,Goodman AL,Kagan JC
Tue Nov 17 00:00:00 EST 2015
Role of caveolin-3 in lymphocyte activation.
Life sciences
Tran C,Stary CM,Schilling JM,Bentley B,Patel HH,Roth DM
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Thu Jan 15 00:00:00 EST 2015
Selective Targeting of a Disease-Related Conformational Isoform of Macrophage Migration Inhibitory Factor Ameliorates Inflammatory Conditions.
Journal of immunology (Baltimore, Md. : 1950)
Thiele M,Kerschbaumer RJ,Tam FW,Völkel D,Douillard P,Schinagl A,Kühnel H,Smith J,McDaid JP,Bhangal G,Yu MC,Pusey CD,Cook HT,Kovarik J,Magelky E,Bhan A,Rieger M,Mudde GC,Ehrlich H,Jilma B,Tilg H,Moschen A,Terhorst C,Scheiflinger F
17-0141 was used in Flow cytometry/Cell sorting to demonstrate that oxMIF represents the disease-related isoform of MIF and is therefore a new diagnostic marker for inflammation.
Tue Sep 01 00:00:00 EDT 2015
Yersinia pestis targets neutrophils via complement receptor 3.
Cellular microbiology
Merritt PM,Nero T,Bohman L,Felek S,Krukonis ES,Marketon MM
17-0141 was used in Flow cytometry/Cell sorting to identify the factors involved in the targeting of murine splenocytes by Yersinia pestis.
Fri May 01 00:00:00 EDT 2015
Lack of CD47 on donor hepatocytes promotes innate immune cell activation and graft loss: a potential barrier to hepatocyte xenotransplantation.
Cell transplantation
Navarro-Alvarez N,Yang YG
17-0141 was used in Flow cytometry/Cell sorting to investigate whether CD47 incompatibility also induces rejection of nonhematopoietic cellular xenografts.
Sat Mar 01 00:00:00 EST 2014
Characterization of cardiac-resident progenitor cells expressing high aldehyde dehydrogenase activity.
BioMed research international
Roehrich ME,Spicher A,Milano G,Vassalli G
17-0141 was used in Flow cytometry/Cell sorting to investigate the presence of cardiac-resident progenitor cells with high aldehyde dehydrogenase activity isolated from mouse hearts.
Tue Sep 03 00:00:00 EDT 2013
Circadian mechanisms in murine and human bone marrow mesenchymal stem cells following dexamethasone exposure.
Bone
Wu X,Yu G,Parks H,Hebert T,Goh BC,Dietrich MA,Pelled G,Izadpanah R,Gazit D,Bunnell BA,Gimble JM
Published figure using CD14 monoclonal antibody (Product # 17-0141-82) in Flow Cytometry
Thu May 01 00:00:00 EDT 2008