Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AYYQQLNTEL KENLKDTMTK RYHQPGHEAV TSAVDQLQQE FHCCGSNNSQ DWRDSEWIRS QEAGGRVVPD SCCKTVVALC GQRDHASNIY KVEGGCITKL ETFIQEHLR
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It is involved in cellular processes including cell adhesion and may regulate integrin trafficking and/or function. This protein enhances cell motility, invasion and metastasis of cancer cells. Multiple alternatively spliced transcript variants that encode the same protein have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD 151; CD151; CD151 antigen; CD151 antigen (Raph blood group); CD151 molecule; GP27; hemidesmosomal tetraspanin CD151; Membrane glycoprotein SFA-1; PETA-3; platelet endothelial tetraspan antigen-3; platelet surface glycoprotein gp27; platelet-endothelial cell tetraspan antigen 3; Platelet-endothelial tetraspan antigen 3; Tetraspanin-24; Tspan-24
Gene Aliases: CD151; GP27; MER2; PETA-3; Peta3; RAPH; SFA-1; SFA1; TSPAN24
UniProt ID: (Human) P48509, (Rat) Q9QZA6, (Mouse) O35566
Entrez Gene ID: (Human) 977, (Rat) 64315, (Mouse) 12476
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support