Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects both GPI-anchored and transmembrane form of CD160.
CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules. CD160 is expressed at the cell surface as a tightly disulfide-linked multimer. It is involved in cell proliferation, cell surface receptor signaling pathways, cellular defense response, and regulation of immune response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD160; CD160 antigen; CD160 transmembrane isoform; CD160-delta Ig; natural killer cell receptor; Natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member
Gene Aliases: BY55; CD160; NK1; NK28
UniProt ID: (Human) O95971
Entrez Gene ID: (Human) 11126
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support