Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Rat anti Mouse CD169 antibody, clone 3D6.112 recognizes mouse CD169 also known as sialoadhesin, Sheep erythrocyte receptor or Siglec-1.
The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD169; SER; Sheep erythrocyte receptor; Sialic acid-binding Ig-like lectin 1; Sialoadhesin; Siglec-1
Gene Aliases: Cd169; Sa; Siglec-1; Siglec1; Sn
UniProt ID: (Mouse) Q62230
Entrez Gene ID: (Mouse) 20612
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support