Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MSPVLLLALL GFILPLPGVQ ALLCQFGTVQ HVWKVSDLPR QWTPKNTSCD SGLGCQDTLM LIESGPQVSL VLSKGCTEAK DQEPRVTEHR MGPGLSLISY TFVCRQEDFC NNLVNSLPLW APQPPADPGS LRCPVCLSME GCLEGTTEEI CPKGTTHCYD GLLRLRGGGI FSNLRVQGCM PQPGCNLLNG TQEIGPVGMT ENCNRKDFLT CHRGTTIMTH GNLAQEPTDW TTSNTEMCEV GQVCQETLLL LDVGLTSTLV GTKGCSTVGA QNSQKTTIHS APPGVLVASY THFCSSDLCN SASSSSVLLN SLPPQAAPVP GDRQCPTCVQ PLGTCSSGSP RMTCPRGTTH CYDGYIHLSG GGLSTKMSIQ GCVAQPSSFL LNHTRQIGIF SAREKRDVQP PASQHEGGGA EGLESLTWGV GLALAPALWW GVVCPSC
CD177 (NB1/HNA-2a and PRV-1 form) is a GPI-anchored glycoprotein present mainly on neutrophils. Its plasma membrane expression is increased during pregnancy and and inflammation or after G-CSF application. Ligand of CD177 has been identified as CD31 (PECAM-1). CD177 participates in neutrophil transmigration and seems to be also a pro-proliferative molecule. The antibodies against CD177 can be involved in neonatal alloimmune neutropenia (NAN).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD177; CD177 antigen; cell surface receptor; HNA-2a; Human neutrophil alloantigen 2a; NB1 glycoprotein; NB1 GP; Polycythemia rubra vera protein 1; PRV-1
Gene Aliases: CD177; HNA-2a; HNA2A; NB1; NB1 GP; PRV-1; PRV1; UNQ595/PRO1181
UniProt ID: (Human) Q8N6Q3
Entrez Gene ID: (Human) 57126
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support