Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: EHAFQGPTSF HVIQTSSFTN STWAQTQGSG WLDDLQIHGW DSDSGTAIFL KPWSKGNFSD KEVAELEEIF RVYIFGFARE VQDFAGDFQM KY
This gene encodes a member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by this gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail, and requires vesicular acidification to bind lipid antigens.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BDCA1; CD1b; CD1B antigen, b polypeptide; CD1C antigen; cortical thymocyte antigen CD1B; differentiation antigen CD1-alpha-3; MGC125990; MGC125991; RP11-101J8.3; RP11-101J8.4; T-cell surface glycoprotein CD1b
Gene Aliases: CD1; CD1A; CD1B; R1
UniProt ID: (Human) P29016
Entrez Gene ID: (Human) 910
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support