Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 4
Description: The AISB12 monoclonal antibody reacts with mouse CD20. CD20 is a B cell-specific 32kDa membrane protein involved in intracellular calcium regulation. CD20 expression begins after the induction of CD19 during development, and is coexpressed with IgM during the pre-B to immature B cell transition in bone marrow. Expression then increases during maturation with almost all mature cells expressing some level of CD20. The human CD20 antigen has proven to be an effective immunotherapeutic target in B cell lymphoma and several autoimmune diseases including rheumatoid arthritis and type I diabetes. CD20 consists of 4 transmembrane domains and a 44 amino acid extracellular loop domain where AISB12 binds.
Applications Reported: This AISB12 antibody has been reported for use in flow cytometric analysis and western blotting.
Applications Tested: This AISB12 antibody has been tested by immunoblot on A20 cell lysates and flow cytometry on mouse splenocytes.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
CD20 is a non-glycosylated surface phosphoprotein that has a molecular weight range of 33-37 kDa depending on the degree of phosphorylation. CD20 is expressed on mature and most malignant B cells, in a subpopulation of T lymphocytes and follicular dendritic cells. CD20 expression on B cells is synchronous with the expression of surface IgM and it regulates transmembrane calcium conductance, cell cycle progression and B-cell proliferation. CD20 is also associated with lipid rafts, but the intensity of this association depends on extracellular triggering, employing CD20 conformational change, and/or BCR (B cell antigen receptor) aggregation. After the receptor ligation, BCR and CD20 colocalize and then rapidly dissociate before BCR endocytosis, whereas CD20 remains at the cell surface. CD20 serves as a useful target for antibody-mediated therapeutic depletion of B cells, as it is expressed at high levels on most B-cell malignancies, but does not become internalized or shed from the plasma membrane following monoclonal antibody treatment. Diseases associated with CD20 dysfunction include Ms4a1-related common variable immune deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APY; ATOPY; B-cell differentiation antigen Ly-44; B-lymphocyte antigen CD20; CD20; CD20 antigen; Fc epsilon receptor I beta chain; Fc Fragment of IgE high affinity I receptor for beta polypeptide; FCER1B; IGEL; IGER; IGHER; LEU16; Ly44; Lymphocyte antigen 44; Membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 2; MGC3969
Gene Aliases: AA960661; Cd20; Ly-44; Ms4a1; Ms4a2
UniProt ID: (Mouse) P19437
Entrez Gene ID: (Rat) 309217, (Mouse) 12482
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support