Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Leinco Technologies
Specificity: Clone 22D1 recognizes an epitope on mouse CD209b.
Endotoxin Level: ≤ 1.0 EU/mg as determined by the LAL method. Purity: >95% by SDS Page; ≥95% monomer by analytical SEC.
Product Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using only In vitro protein free cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Storage and Handling: Functional grade preclinical antibodies may be stored sterile as received at 2-8 C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80 C. Avoid Repeated Freeze Thaw Cycles.
This antibody has been tested in the following applications: Western Blotting, Immunohistochemistry-Fresh Frozen, Flow Cytometry, Blocking.
SIGNR1 is a type II transmembrane C-type lectin that was identified in a search for mouse homologues of human DC-SIGN. It is expressed at high levels in splenic marginal zone macrophages and lymph node medullary macrophages, where it functions to uptake dextran polysaccharides, including the capsular polysaccharide of Streptococcus pneumoniae. It has also been demonstrated that SIGNR1 physically associates with TLR4/MD2, and it has been suggested that this association plays a role in recognition of LPS. Furthermore, recently it has been shown that SIGNR1 deficient mice have a defect in catabolism of the complement component C3, and that SIGNR1 binds directly to the complement C1 subcomponent, C1q to assemble a non-conventional C3 convertase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD209; CD209 antigen-like protein A; CD209 antigen-like protein B; DC SIGNR1; DC-SIGN; DC-SIGN-related protein 1; DC-SIGNR1; Dendritic cell-specific ICAM-3-grabbing non-integrin; MGC130443; OtB7
Gene Aliases: 1810030I22Rik; CD209; Cd209a; Cd209b; CDSIGN; CIRE; DC-SIGN; DC-SIGN1; DC-SIGNR1; Dcsign; mSIGNR1; OtB7; SIGN-R1; SIGNR1; SIGNR5
UniProt ID: (Mouse) Q91ZX1, (Mouse) Q8CJ91
Entrez Gene ID: (Mouse) 170786, (Mouse) 69165
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support