Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: The H44 antibody reacts with the interleukin-18 receptor alpha chain, also known as IL-1Rrp, an IL-1R family member. IL-18R alpha is expressed on Th1 cells, NK cell subset, neutrophils and IL-12-stimulated tonsillar B cells. By itself, IL-18R alpha binds IL-18 with low affinity and when complexed with IL-18R beta chain binds IL-18 with high affinity. The proinflammatory cytokine IL-18 and its receptor complex play an important role in the innate and autoimmune responses.
Applications Reported: H44 has been reported for use in flow cytometric analysis.
Applications Tested: This H44 antibody has been pre-titrated and tested by flow cytometric analysis on human peripheral blood cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
It is recommended to stain whole blood before RBC lysis.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Filtration: 0.2 µm post-manufacturing filtered.
IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL-18R-1; IL-18R-alpha; IL-18R1; IL-1Rrp; IL1 receptor-related protein; IL18Ralpha2; IL1R-rp; Interleukin-18 receptor 1; Interleukin-18 receptor alpha
Gene Aliases: CD218a; CDw218a; IL-1Rrp; IL18R1; IL18RA; IL1RRP
UniProt ID: (Human) Q13478
Entrez Gene ID: (Human) 8809
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support