Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDYPTLLLA LLHVYRALCE EVLWHTSVPF AENMSLECVY PSMGILTQVE WFKIGTQQDS IAIFSPTHGM VIRKPYAERV YFLNSTMASN NMTLFFRNAS EDDVGYYSCS LYTYPQGTWQ KVIQVVQSDS FEAAVPSNSH IVSEPGKNVT LTCQPQMTWP VQAVRWEKIQ PRQIDLLTYC NLVHGRNFTS KFPRQIVSNC SHGRWSVIVI PDVTVSDSGL YRCYLQASAG ENETFVMRLT VAEGKTDNQY TLFVA (1-254 aa encoded by BC074787)
DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adhesion glycoprotein; CD226; CD226 antigen; DNAM 1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; T lineage-specific activation antigen 1 antigen
Gene Aliases: CD226; DNAM-1; DNAM1; PTA1; TLiSA1
UniProt ID: (Human) Q15762
Entrez Gene ID: (Human) 10666
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support