Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Description: This 10E5 monoclonal antibody reacts with mouse CD226, which is also known as DNAX accessory molecule-1 (DNAM-1). CD226 is an adhesion molecule expressed primarily on T cells, including thymocytes. The highest levels of CD226 are found on naive CD8 T cells. Moreover, expression of CD226 is regulated differentially in CD4 T cells. Under certain culture conditions, CD226 can be upregulated in Th1 and downregulated in Th2 and Th0 cells. This cell surface receptor can also be detected on natural killer cells, some B cell subsets, monocytes, and platelets. Interaction with its ligands, the poliovirus receptor (CD155) and CD112, results in adhesion and cell signaling events that promote innate and adaptive immunity, including differentiation, survival, and cytotoxicity.
The 10E5 monoclonal antibody has been reported to inhibit antigen-specific T cell proliferation and IFN-gamma production in vivo.
Applications Reported: This 10E5 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This 10E5 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
Filtration: 0.2 µm post-manufacturing filtered.
DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: CD226; CD226 antigen; DNAM 1; platelet and T cell activation antigen 1; Platelet and T-cell activation antigen 1
Gene Aliases: BC051526; Cd226; DNAM-1; DNAM1; Pta1; TLiSA1
UniProt ID: (Mouse) Q8K4F0
Entrez Gene ID: (Mouse) 225825
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support