Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Specificity: This antibody is specific for murine CD226.
Preparation: This antibody was raised by immunising Lewis and Lou/M rats with the murine Th1 cell clone AE7 and Th1 cell lines derived from 5B6 and DO11.10 TCR transgenic mice, before screening for reactivity with Th1 clones and a lack of reactivity to Th2/Th0 cell lines.
DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD226; CD226 antigen; DNAM 1; platelet and T cell activation antigen 1; Platelet and T-cell activation antigen 1
Gene Aliases: BC051526; Cd226; DNAM-1; DNAM1; Pta1; TLiSA1
UniProt ID: (Mouse) Q8K4F0
Entrez Gene ID: (Mouse) 225825
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support