Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
BTLA or B and T-lymphocyte attenuator is a member of the co-inhibitory receptors of the CD28 superfamily. BTLA is a lymphocyte inhibitory receptor which inhibits lymphocytes during immunes responses. BTLA is constitutively expressed on most CD4+ and CD8+ T cells and its expression progressively decreases upon T cell activation. It remains expressed on Th1 cells, but not Th2 cells. BTLA is a unique co-receptor that interacts with the tumor necrosis factor receptor superfamily member herpesvirus entry mediator (HVEM). This interation is an important pathway regulating lymphocyte activation and/or homeostasis in the immune response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B and T lymphocyte associated protein; B and T lymphocyte attenuator; B- and T-lymphocyte attenuator; B- and T-lymphocyte-associated protein; BTLA_HUMAN; CD272; CD272 antigen; FLJ16065; MGC129743
Gene Aliases: A630002H24; BTLA; BTLA1; CD272
UniProt ID: (Human) Q7Z6A9, (Rat) Q6PNM1, (Mouse) Q7TSA3
Entrez Gene ID: (Human) 151888, (Rat) 407756, (Mouse) 208154
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support