Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: 293T, U-87MG, Mouse lung, Mouse testis, Mouse brain, Rat lung, Rat testis, Rat spleen
Immunogen sequence: GLKPEGVPRI DCVSKNEDIP NVAVYPHNGM IDLKYFPYYG KKLHVGYLQP LVAVQVSFAP NNTGKEVTVE CKIDGSANLK SQDDRDKFLG RVMFKITARA
The protein encoded by this gene belongs to the family of Na+/K+ and H+/K+ ATPases beta chain proteins, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane. The glycoprotein subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes a beta 3 subunit. This gene encodes a beta 3 subunit. A pseudogene exists for this gene, and it is located on chromosome 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, Na+/K+ beta 3 polypeptide; ATPase, Na+K+ transporting, beta polypeptide 3; ATPB-3; CD298; CD298 antigen; Na K-ATPase beta-3 subunit; Na+/K+ -ATPase beta 3 subunit; Sodium Potassium ATPase; Sodium/potassium-dependent ATPase subunit beta-3; Sodium/potassium-transporting ATPase subunit beta-3
Gene Aliases: AA409958; AI664000; ATP1B3; ATPB-3; AW212096; CD298; NKAB3S
UniProt ID: (Human) P54709, (Mouse) P97370, (Rat) Q63377
Entrez Gene ID: (Human) 483, (Mouse) 11933, (Rat) 25390
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support