Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody recognizes an extracellular epitope on CD300e / IREM-2, a 32 kDa glycoprotein expressed by mature monocytes and peripheral blood myeloid dendritic cells.
Functional application: Stimulation
CD300e is a member of the glycoprotein family of cell surface proteins expressed on myeloid cells. CD300e interacts with the TYRO protein tyrosine kinase-binding protein and is thought to be involved as an activating receptor. CD300e is expressed on mature myeloid dendritic cells and monocytes, and is down regulated on immature dendritic cells. Further, CD300e interacts with DAP-12, which mediates activating signals. The alloreactive response of naïve T cells has been reported to become enhanced by the activation of CD300e. Diseases associated with CD300e dysfunction include tungiasis and lice infestation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD300 antigen like family member E; CD300 antigen-like family member E; CD300e; CD300e antigen; CLM-2; CMRF35-A5; CMRF35-like molecule 2; Immune receptor expressed on myeloid cells 2; IREM-2; PIgR-2; poly-Ig receptor 2; Polymeric immunoglobulin receptor 2
Gene Aliases: CD300E; CD300LE; CLM-2; CLM2; CMRF35-A5; CMRF35A5; IREM-2; IREM2; PIgR-2; PIgR2
UniProt ID: (Human) Q496F6
Entrez Gene ID: (Human) 342510
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support