Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: The UP-H2 monoclonal antibody recognizes human IREM-2 (immune receptor expressed in monocytic derived cells) also known as LMIR6, and unofficially CD300E and CD300LE. It is a member of the CMRF-35 family. This 34 kDa Ig-like containing receptor is an activating receptor and through it's charged residues in the cytokplasmic domain interacts with ITAM containing adaptor proteins like DAP12. Expression is found exclusively on monocytes and myeloid DC's. Expression may be transient as differentiation of monocytes leads to down regulation of IREM-2 yet expression is found on mature DCs. It has been also noted that myeloid cell lines such as THP-1 and U937 do not express IREM-2.
In vitro addition of UP-H2 antibody either soluble or crosslinked stimulates monocytes.
Applications Reported: This UP-H2 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This UP-H2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
Filtration: 0.2 µm post-manufacturing filtered.
CD300e is a member of the glycoprotein family of cell surface proteins expressed on myeloid cells. CD300e interacts with the TYRO protein tyrosine kinase-binding protein and is thought to be involved as an activating receptor. CD300e is expressed on mature myeloid dendritic cells and monocytes, and is down regulated on immature dendritic cells. Further, CD300e interacts with DAP-12, which mediates activating signals. The alloreactive response of naïve T cells has been reported to become enhanced by the activation of CD300e. Diseases associated with CD300e dysfunction include tungiasis and lice infestation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD300 antigen like family member E; CD300 antigen-like family member E; CD300e; CD300e antigen; CLM-2; CMRF35-A5; CMRF35-like molecule 2; Immune receptor expressed on myeloid cells 2; IREM-2; PIgR-2; poly-Ig receptor 2; Polymeric immunoglobulin receptor 2
Gene Aliases: CD300E; CD300LE; CLM-2; CLM2; CMRF35-A5; CMRF35A5; IREM-2; IREM2; PIgR-2; PIgR2
UniProt ID: (Human) Q496F6
Entrez Gene ID: (Human) 342510
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support