Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: CD305 or Leukocyte-associated Ig-like receptor-1 (LAIR-1) is a 46 kD cell surface molecule that functions as an inhibitory collagen-receptor on the majority of cells of the immune system. The mouse LAIR-1 molecule is expressed primarily on monocytes/macrophages and also on T cells, NK cells and dendritic cells. Unlike human B cells , mouse B cells do not express LAIR-1. The cytoplasmic tail of mouse LAIR-1 can be phosphorylated leading to the recruitment of SH2-containing tyrosine phosphatase(SHP)-2 and C-terminal Src kinase.
Applications Reported: This 113 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunoblotting (WB). It has also been reported for use in functional studies.
Applications Tested: This 113 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Storage and handling: Use in a sterile environment.
Filtration: 0.2 µm post-manufacturing filtered.
Purity: Greater than 90%, as determined by SDS-PAGE.
Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.
Aggregation: Less than 10%, as determined by HPLC.
LAIR1 (CD305) functions as an inhibitory receptor that plays a constitutive negative regulatory role on cytolytic function of natural killer (NK) cells, B-cells and T-cells. Activation by Tyr phosphorylation results in recruitment and activation of the phosphatases PTPN6 and PTPN11. It also reduces the increase of intracellular calcium evoked by B-cell receptor ligation. May also play its inhibitory role independently of SH2-containing phosphatases. Modulates cytokine production in CD4+ T-cells, down-regulating IL2 and IFNG production while inducing secretion of transforming growth factor beta. Down-regulates also IgG and IgE production in B-cells as well as IL8, IL10 and TNF secretion. Inhibits proliferation and induces apoptosis in myeloid leukemia cell lines as well as prevents nuclear translocation of NF-kappa-B p65 subunit/RELA and phosphorylation of I-kappa-B alpha/CHUK in these cells. Inhibits the differentiation of peripheral blood precursors towards dendritic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD305; LAIR-1; Leukocyte-associated immunoglobulin-like receptor 1
Gene Aliases: 5133400O11Rik; BB115266; D7Bwg0421e; Lair-1; Lair1
UniProt ID: (Mouse) Q8BG84
Entrez Gene ID: (Mouse) 52855
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support