Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Mouse anti Bovine CD32 antibody, clone CCG36 recognizes the bovine homologue of human CD32, one of a group of Fc receptors belonging to the immunoglobulin superfamily and involved in phagocytosis of opsonized microbes.
Reconstitute with 1.0 mL distilled water.
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD32; CD32 receptor 2; CD32 receptor 2a; Fc fragment of IgG, low affinity IIa, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIc, receptor for (CD32); Fc-gamma RII; Fc-gamma-RII-D; FcRII; FGFR2B; IgG Fc receptor II; Low affinity immunoglobulin gamma Fc region receptor II; MGC23887; MGC30032; RP11-5K23.6; soluble receptor; boFcRII-2
Gene Aliases: CD32; FCGR2; FCGR2C; fcRII
UniProt ID: (Bovine) Q28110
Entrez Gene ID: (Bovine) 782652, (Bovine) 282229
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support