Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody recognizes a cell surface antigen of 40-42 kDa which is similar to the human cell surface antigen designated as CD32. It has therefore been hypothesized that clone VPM63 may recognize a minor isoform of the ovine homologue of human CD32.
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD32 receptor 2; CD32 receptor 2a; FGFR2B; IgG Fc gamma receptor II; MGC23887; MGC30032; RP11-5K23.6
Entrez Gene ID: (Sheep) 100192432
Molecular Function: immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support