Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: The monoclonal antibody AT130-2 reacts with mouse CD32b, also known as Fc-gamma receptor IIb. CD32b is the only inhibitory member of the Fc-gamma receptor family. Immune complex engagement of CD32b results in the phosphorylation of its intracellular ITIM motif. This engagement serves to attenuate B cell receptor (BCR) signaling and other activating signals, reduces proliferation, reduces antibody production, and may also deliver apoptotic signals. CD32b is the only Fc-gamma receptor expressed by B cells. CD32b is also expressed on monocytes, dendritic cells, neutrophils, eosinophils and basophils, as well as B cell progenitors and plasma cells.
AT130-2 has been reported to have agonist activity.
Applications Reported: This AT130-2 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This AT130-2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
Filtration: 0.2 µm post-manufacturing filtered.
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD32; CD32 receptor 2; CD32 receptor 2a; CD32 receptor 2b; CD32A; Fc gamma receptor IIB; Fc gamma RIIB; Fc-gamma RII; fc-gamma RII-a; Fc-gamma-RII; fc-gamma-RIIa; fc-gamma-RIIB; FcgammaRIIB; FcGR; FCGR2A1; fcRII-a; FGFR2B; IgG Fc receptor II beta; Low affinity immunoglobulin gamma Fc region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; Ly-17; Lymphocyte antigen 17
Gene Aliases: AI528646; CD32; F630109E10Rik; Fc[g]RII; Fcgr2; Fcgr2a; Fcgr2b; FcgRII; Fcr-2; Fcr-3; fcRII; Ly-17; Ly-m20; LyM-1
UniProt ID: (Mouse) P08101
Entrez Gene ID: (Mouse) 14130
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support