Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Specificity: This antibody is specific for murine GITR, a member of the tumor necrosis factor receptor (TNFR) family that is expressed at low levels on unstimulated T cells, B cells, and macrophages. Upon activation, CD4+ and CD8+ T cells up-regulate GITR expression, whereas immunoregulatory T cells constitutively express high levels of GITR.
Preparation: This antibody was raised by immunising Wistar rats with the CD25+CD4+ T cell line. Hybridoma cells were generated by fusing spleen cells from the immunized rat with P3X63Ag8.653 myeloma cells, assessed for secreting antibodies capable of neutralizing in vitro CD25+CD4+ T cell-mediated suppression of anti-CD3-stimulated T cell proliferation9, and then cloned. This DTA-1 clone secreted a monoclonal rat immunoglobulin G2a (IgG2a) that showed high neutralizing activity.
Tumor necrosis factor receptor superfamily member 18 (TNFRSF18), also called GITR or AITR is a protein that in humans is encoded by the TNFRSF18 gene. It is mapped to 1p36.33. This gene encodes a member of the TNF-receptor superfamily. The encoded receptor has been shown to have increased expression upon T-cell activation, and it is thought to play a key role in dominant immunological self-tolerance maintained by CD25 (+)CD4 (+) regulatory T cells. Knockout studies in mice also suggest the role of this receptor is in the regulation of CD3-driven T-cell activation and programmed cell death. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD357; Glucocorticoid-induced TNFR-related protein; PRO364; Tumor necrosis factor receptor superfamily member 18; UNQ319
Gene Aliases: AITR; Gitr; Tnfrsf18
UniProt ID: (Mouse) O35714
Entrez Gene ID: (Mouse) 21936
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support