Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Single-chain membrane glycoprotein that forms a non-covalent complex with GPIb. (MW 23 kDa) Reactivity with resting and activated platelets, weakly on monocytes, megakaryocytes and attachment site for the platelet plasma membrane to the submembrane cytoskeleton. GPIb/IX complex, functions as the receptor for ristocetin-induced binding of von Willebrand factor and as the von Willebrand factor-depend adhesion receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD42a; Glycoprotein 9; glycoprotein IX (platelet); GP-IX; Platelet glycoprotein IX; Platelet GPIX
Gene Aliases: Cd42; CD42a; GP9; GPIX
UniProt ID: (Human) P14770, (Mouse) O88186
Entrez Gene ID: (Human) 2815, (Mouse) 54368
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support