Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KMSSVPQET PHATSHPAVP ITANSLGSHT VTGGTITTNS PETSSRTSGA PVTTAASSLE TSRGTSGPPL TMATVSLETS KGTSGPPVTM ATDSLETSTG TTGPPVTMTT GSLEPSSGAS GPQVSSVKLS TMMSPTTSTN ASTVPFRNPD ENSRGMLPVA VLVALLAVIV LVALLLLWRR RQKRRTGALV LSRGGKRNGV VDAWAGPAQV PEEGAVTVTV GGSGGDKGSG FPDGEGSSRR PTLTTFFGRR KSRQGSLAME ELKSGSGPSL KGEEEPLVAS EDGAVDAPAP DEPEGGDGAA P (101-400 aa encoded by BC012350)
CD43 (leukosialin, sialophorin) is a transmembrane mucin-like protein with high negative charge, expressed on the surface of most hematopoietic cells. CD43 contributes to a repulsive barrier that interferes with cellular adhesion, however, in certain cases also promotes leukocyte aggregation. By interaction with actin-binding proteins ezrin and moesin CD43 plays a regulatory role in remodeling T-cell morphology and regulates cell-cell interactions during lymphocyte traffic. CD43 signaling both enhances LFA-1 adhesiveness and counteracts LFA-1 induction via other receptors. Expression of CD43 causes induction of functionally active tumor suppressor p53 protein, but in case of p53 and ARF deficiency CD43 promotes tumor proliferation and viability. CD43 appears to be an important modulator of leukocyte functions. Diseases associated with CD43 dysfunction include Wiscott-Aldrich Syndrome and Adenoid Basal Cell Carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD43; Galactoglycoprotein; GALGP; GPL115; Leukocyte sialoglycoprotein; Leukosialin; Sialophorin; sialophorin (gpL115, leukosialin, CD43); sialophorin gpL115
Gene Aliases: CD43; GALGP; GPL115; LSN; SPN
UniProt ID: (Human) P16150
Entrez Gene ID: (Human) 6693
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support