Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute by adding 1 mL distilled water.
CD45RC (PTPRC) is a protein tyrosine-protein phosphatase required for T-cell activation through the antigen receptor. It acts as a positive regulator of T-cell coactivation upon binding to DPP4. The first PTPase domain has enzymatic activity, the second affects the substrate specificity of the first. Upon T-cell activation, CD45RC recruits and dephosphorylates SKAP1 and FYN. Mutations of the gene can result in severe combined immunodeficiency and multiple sclerosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD45 antigen; protein tyrosine phosphatase; alternatively spliced; receptor-type tyrosine-protein phosphatase C
Gene Aliases: CD45
Entrez Gene ID: (Pig) 100522631
Molecular Function:
extracellular matrix protein
protein phosphatase
transmembrane signal receptor
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support