Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for Human CD47.
CD47 (integrin-associated protein, IAP) is a ubiquitously expressed cell surface transmembrane glycoprotein interacting with several integrins and regulating their functions. Engagement of CD47 by soluble ligands or counter receptors modulates various signaling pathways, such as activation of heterotrimeric G proteins. Functionally, CD47 is involved in the increase in intracellular calcium concentration that occurs upon cell adhesion to extracellular matrix. CD47 is also a receptor for the C-terminal cell binding domain of thrombospondin, and it may play a role in membrane transport and signal transduction. CD47 has broad tissue distribution, is reduced in expression on Rh erythrocytes. CD47 acts as a ligand for CD172a (signal regulatory protein alpha, SIRP alpha), an immune inhibitory receptor on macrophages; this interaction prevents phagocytosis of CD47-positive cells. Moreover, CD47-CD172a system affects cell migration, B cell adhesion, T cell activation, and is involved in modulation of chondrocyte responses to mechanical signals, and promotes neuronal development, being especially abundant in synapse-rich regions of brain and retina.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen identified by monoclonal antibody 1D8; Antigenic surface determinant protein OA3; CD47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 glycoprotein; IAP; integrin associated protein; Integrin-associated protein; integrin-associated signal transducer; Leukocyte surface antigen CD47; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47
Gene Aliases: CD47; IAP; MER6; OA3
UniProt ID: (Human) Q08722
Entrez Gene ID: (Human) 961
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support