Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD48 (Met1-Arg216) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze!
CD48 (Blast-1) belongs to the CD2 subset of the Ig superfamily that includes CD2, CD2F-10, CD58, CD84, CD150, CD229, CD244 and others. These molecules bind to the same or another members of the Ig family, thus mediating homotypic or heterotypic adhesion. CD48 is a GPI-anchored protein broadly expressed on hematopoietic cells and serves as a high affinity ligand for 2B4 and low affinity ligand for CD2. 2B4-CD48 interaction among NK cells and NK-T cells regulates cell proliferation. Further, signaling through CD48 results in eosinophil activation and CD48 expression is increased in several infectious diseases. CD48 is strongly expressed on lymphocytes and monocytes, weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of marker for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2. 2B4-CD48 interaction may be involved in the pathology of allergic airway diseases and provide a pharmacological target for the treatment of diseases such as allergic rhinitis and asthma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell activation marker; BCM1 surface antigen; BLAST-1; CD48; CD48 antigen; CD48 antigen (B cell membrane protein); CD48d; MRC OX-45 surface antigen; sCD 48; sCD48; Signaling lymphocytic activation molecule 2; Signaling lymphocytic activation molecule 2 (SLAMF2); SLAM family member 2; SLAMF2; soluble CD 48; soluble CD48
Gene Aliases: Bcm-1; Cd48; SLAMF2
UniProt ID: (Rat) P10252
Entrez Gene ID: (Rat) 245962
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support