Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Immunogen Sequence: FNLDAEAPAV LSGPPGSFFG FSVEFYRPGT DGVSVLVGAP KANTSQPGVL QGGAVYLCPW GASPTQCTPI EFDSKGSRLL ESSLSSSEGE EPVEYKSLQW FGATVRAHGS SILACAPLYS WRTEKEPLSD PVGTCYLSTD NFTRILEYAP CRSDFSWAAG QGYCQGGFSA EFTKTGRVVL GGPGSYFWQG QILSATQEQI AESYYPEYLI NLVQGQLQTR QASSIYDDSY LGYSVAVGEF SGDDTEDFVA GVPKGNLTYG YVTILNGSDI RSLYNFSGEQ MASYFGYAVA ATDVNGDGLD DLLVGAPLLM DRTPDGRPQE VGRVYVYLQH PAGIEPTPTL TLTGHDEFGR FGSSLTPLGD LDQDGYNDVA IGAPFGGETQ QGVVFVFPGG PGGLGSKPSQ VLQPLWAASH TPDFFGSALR GGRDLDGNGY PDLIVGSFGV DKAVVYRGRP IVSASASLTI FPAMFNPEER SCSLEGNPVA CINLSFCLNA SGKHVADSIG FTVELQLDWQ KQKGGVRRAL FLASRQATLT QTLLIQNGAR EDCREMKIYL RNESEFRDKL SPIHIALNFS LDPQAPVDSH GLRPALHYQS KSRIEDKAQI LLDCGEDNIC VPDLQLEVFG EQNHVYLGDK NALNLTFHAQ NVGEGGAYEA ELRVTAPPEA EYSGLVRHPG NFSSLSCDYF AVNQSRLLVC DLGNPMKAGA SLWGGLRFTV PHLRDTKKTI QFDFQILSKN LNNSQSDVVS FRLSVEAQAQ VTLNGVSKPE AVLFPVSDWH PRDQPQKEED LGPAVHHVYE LINQGPSSIS QGVLELSCPQ ALEGQQLLYV TRVTGLNCTT NHPINPKGLE LDPEGSLHHQ QKREAPSRSS ASSGPQILKC PEAECFRLRC ELGPLHQQES QSLQLHFRVW AKTFLQREHQ PFSLQCEAVY KALKMPYRIL PRQLPQKERQ VATAVQWTKA EGSY
The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD49 antigen-like family member E; CD49e; Fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; integrin alpha 5; Integrin alpha-5; Integrin alpha-F; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); very late activation protein 5, alpha subunit; VLA-5
Gene Aliases: CD49e; FNRA; ITGA5; VLA-5; VLA5A
UniProt ID: (Human) P08648
Entrez Gene ID: (Human) 3678
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support