Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: The eBioHMalpha5-1 monoclonal antibody reacts with mouse and rat CD49e, also known as integrin alpha5 or VLA-5A. CD49e is a member of the integrin alpha chain family, and binds with CD29 (integrin beta1) to form the receptor VLA-5, which binds the ligands fibronectin and fibrinogen. CD49e is a type I, single-pass membrane protein, with a molecular weight of 135 kDa, and is expressed on multiple cell types including thymocytes, mast cells, activated T cells and splenic B cells. In addition to its role in adhesion, VLA-5 contributes to T-cell co-stimulation. The eBioHMalpha5-1 monoclonal antibody has been demonstrated to interfere with VLA-5-mediated cell adhesion, and with the degranulation of IgE-sensitized rat RBL cells stimulated on fibronectin-coated plates. Furthermore, HMalpha5-1 injected into mice subcutaneously, along with anti-CD49d and anti-CD61, is able to inhibit passive cutaneous anaphylaxis.
Applications Reported: This eBioHMa5-1 (HMa5-1) antibody has been reported for use in flow cytometric analysis.
Applications Tested: This eBioHMa5-1 (HMa5-1) antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.
Filtration: 0.2 µm post-manufacturing filtered.
The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD49 antigen-like family member E; CD49e; fibronectin receptor alpha polypeptide; Fibronectin receptor subunit alpha; integrin alpha 5 (fibronectin receptor alpha); Integrin alpha-5; Integrin alpha-F; VLA-5
Gene Aliases: Cd49e; Fnra; Itga5; Itga5_retired; VLA5
UniProt ID: (Mouse) P11688
Entrez Gene ID: (Mouse) 16402, (Rat) 315346
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support