Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
CD51 and CD61 molecules (alpha V and beta 3 integrins) form a complex. This complex binds vitronection at the RGD sequence and can also bind fibrinogen, von Willebrand factor, thrombospondin, fibronectin, osteopontin and collagen. Reacts with osteoclasts, placenta, melanoma cell lines and weakly with platelets.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen CD61; antigen identified by monoclonal antibody L230; CD51; CD61; GPIIIa; integrin alpha V; integrin alpha-5 beta-3; Integrin alpha-V; integrin alphaVbeta3; integrin beta 3; Integrin beta-3; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); Platelet membrane glycoprotein IIIa; Vitronectin receptor; Vitronectin receptor subunit alpha
Gene Aliases: BDPLT16; BDPLT2; CD51; CD61; GP3A; GPIIIa; GT; ITGAV; ITGB3; MSK8; VNRA; VTNR
UniProt ID: (Human) P06756, (Human) P05106
Entrez Gene ID: (Human) 3685, (Human) 3690
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support