Product References
Differences in Peripheral Blood Lymphocytes between Brand-Name and Generic Tacrolimus Used in Stable Liver Transplant Recipients.
Medical principles and practice : international journal of the Kuwait University, Health Science Centre
Kim JM,Kwon CHD,Joh JW,Sinn DH,Choi GS,Park JB,Kang ES,Lee SK
MA119129 was used in flow cytometry to characterize the periferal blood lymphocytes phenotype in tacrolimus-treated liver transplanted patients.
Mon Mar 19 00:00:00 EDT 2018
Effect of Anti-IL-15 Administration on T Cell and NK Cell Homeostasis in Rhesus Macaques.
Journal of immunology (Baltimore, Md. : 1950)
DeGottardi MQ,Okoye AA,Vaidya M,Talla A,Konfe AL,Reyes MD,Clock JA,Duell DM,Legasse AW,Sabnis A,Park BS,Axthelm MK,Estes JD,Reiman KA,Sekaly RP,Picker LJ
MA119129 was used in flow cytometry to report the effect of IL-15 inhibition on T and natural killer cell dynamics in rhesus macaques
Mon Aug 15 00:00:00 EDT 2016
LECT2 drives haematopoietic stem cell expansion and mobilization via regulating the macrophages and osteolineage cells.
Nature communications
Lu XJ,Chen Q,Rong YJ,Yang GJ,Li CH,Xu NY,Yu CH,Wang HY,Zhang S,Shi YH,Chen J
MA119129 was used in flow cytometry to find that leukocyte cell-derived chemotaxin 2 promotes expansion and mobilization of hematopoietic stem cells
Tue Sep 06 00:00:00 EDT 2016
New IDH1 mutant inhibitors for treatment of acute myeloid leukemia.
Nature chemical biology
Okoye-Okafor UC,Bartholdy B,Cartier J,Gao EN,Pietrak B,Rendina AR,Rominger C,Quinn C,Smallwood A,Wiggall KJ,Reif AJ,Schmidt SJ,Qi H,Zhao H,Joberty G,Faelth-Savitski M,Bantscheff M,Drewes G,Duraiswami C,Brady P,Groy A,Narayanagari SR,Antony-Debre I,Mitchell K,Wang HR,Kao YR,Christopeit M,Carvajal L,Barreyro L,Paietta E,Makishima H,Will B,Concha N,Adams ND,Schwartz B,McCabe MT,Maciejewski J,Verma A,Steidl U
MA1-19129 was used in flow cytometry to assess the effects of allosteric inhibitors on different mutant forms of isocitrate dehydrogenase 1 in leukemia
Sun Nov 01 00:00:00 EDT 2015
Brief Report: IFIH1 Mutation Causes Systemic Lupus Erythematosus With Selective IgA Deficiency.
Arthritis & rheumatology (Hoboken, N.J.)
Van Eyck L,De Somer L,Pombal D,Bornschein S,Frans G,Humblet-Baron S,Moens L,de Zegher F,Bossuyt X,Wouters C,Liston A
MA1-19129 was used in Flow cytometry/Cell sorting to determine the underlying genetic defect responsible for severe early onset and refractory systemic lupus erythematosus.
Mon Jun 01 00:00:00 EDT 2015
Variation in Gamma-Globin Expression before and after Induction with Hydroxyurea Associated with BCL11A, KLF1 and TAL1.
PloS one
Grieco AJ,Billett HH,Green NS,Driscoll MC,Bouhassira EE
MA1-19129 was used in flow cytometry to elucidate regulation of γ-globin expression in F-cells.
Mon Feb 29 00:00:00 EST 2016
Impaired Th17 polarization of phenotypically naive CD4(+) T-cells during chronic HIV-1 infection and potential restoration with early ART.
Retrovirology
DaFonseca S,Niessl J,Pouvreau S,Wacleche VS,Gosselin A,Cleret-Buhot A,Bernard N,Tremblay C,Jenabian MA,Routy JP,Ancuta P
MA119129 was used in flow cytometry to elucidate Th17 cell polarization, depletion, and restoration in response to HIV infection and antiretroviral therapy
Thu Apr 30 00:00:00 EDT 2015
Grb10 deletion enhances muscle cell proliferation, differentiation and GLUT4 plasma membrane translocation.
Journal of cellular physiology
Mokbel N,Hoffman NJ,Girgis CM,Small L,Turner N,Daly RJ,Cooney GJ,Holt LJ
MA1-19129 was used in flow cytometry to study the increased muscle cell differentiation/proliferation and plama membrane translocation of Glut4 in mice deleted for Grb10
Sat Nov 01 00:00:00 EDT 2014
The endogenous danger signals HSP70 and MICA cooperate in the activation of cytotoxic effector functions of NK cells.
Journal of cellular and molecular medicine
Elsner L,Flügge PF,Lozano J,Muppala V,Eiz-Vesper B,Demiroglu SY,Malzahn D,Herrmann T,Brunner E,Bickeböller H,Multhoff G,Walter L,Dressel R
MA1-19129 was used in flow cytometry to report that HSP70 activates human NK cells to kill target cells expressing MICA in a natural killer group 2 member D-dependent manner
Thu Apr 01 00:00:00 EDT 2010
A new protocol for multiple inhalation of IFN-gamma successfully treats MDR-TB: a case study.
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease
Grahmann PR,Braun RK
MA119129 was used in flow cytometry to demonstrate that aerosolized interferon-gamma results in the transient conversion of sputum smears in multidrug-resistant pulmonary tuberculosis
Sun Jun 01 00:00:00 EDT 2008
Adhesion of human hematopoietic progenitor cells to mesenchymal stromal cells involves CD44.
Cells, tissues, organs
Wagner W,Wein F,Roderburg C,Saffrich R,Diehlmann A,Eckstein V,Ho AD
MA1-19129 was used in blocking or activating experiment to test whether CD44 aids in adhesion of human hematopoietic progenitor cells to mesenchymal stromal cells
Fri Sep 19 00:00:00 EDT 2008
Unique characteristics of NK cells throughout the human female reproductive tract.
Clinical immunology (Orlando, Fla.)
Mselle TF,Meadows SK,Eriksson M,Smith JM,Shen L,Wira CR,Sentman CL
MA1-19129 was used in flow cytometry to characterize natural killer cells from different tissues of the FRT
Sun Jul 01 00:00:00 EDT 2007
Isolation of large numbers of mesenchymal stem cells from the washings of bone marrow collection bags: characterization of fresh mesenchymal stem cells.
Transplantation
Mageed AS,Pietryga DW,DeHeer DH,West RA
MA1-19129 was used in flow cytometry to test if the normally discarded bone marrow collection kits are a convenient source of large numbers of mesenchymal stem cells
Fri Apr 27 00:00:00 EDT 2007
TLRs mediate IFN-gamma production by human uterine NK cells in endometrium.
Journal of immunology (Baltimore, Md. : 1950)
Eriksson M,Meadows SK,Basu S,Mselle TF,Wira CR,Sentman CL
MA1-19129 was used in flow cytometry to assess TLR expression and signaling by uterine natural killer cells from the human endometrium
Mon May 15 00:00:00 EDT 2006
Phenotypic and functional differences between human saphenous vein (HSVEC) and umbilical vein (HUVEC) endothelial cells.
Atherosclerosis
Tan PH,Chan C,Xue SA,Dong R,Ananthesayanan B,Manunta M,Kerouedan C,Cheshire NJ,Wolfe JH,Haskard DO,Taylor KM,George AJ
MA1-19129 was used in flow cytometry to compare human saphenous vein endothelial cells with human umbilical vein endothelial cells
Thu Apr 01 00:00:00 EST 2004
Molecular characterization of human 4Ig-B7-H3, a member of the B7 family with four Ig-like domains.
Journal of immunology (Baltimore, Md. : 1950)
Steinberger P,Majdic O,Derdak SV,Pfistershammer K,Kirchberger S,Klauser C,Zlabinger G,Pickl WF,Stöckl J,Knapp W
MA1-19129 was used in flow cytometry to characterize B7-H3.
Sun Feb 15 00:00:00 EST 2004
LIME: a new membrane Raft-associated adaptor protein involved in CD4 and CD8 coreceptor signaling.
The Journal of experimental medicine
Brdicková N,Brdicka T,Angelisová P,Horváth O,Spicka J,Hilgert I,Paces J,Simeoni L,Kliche S,Merten C,Schraven B,Horejsí V
MA1-19129 was used in flow cytometry to describe a new raft-associated adaptor protein, Lck-interacting molecule, expressed predominantly in T lymphocytes.
Mon Nov 17 00:00:00 EST 2003
Trafficking machinery of NKT cells: shared and differential chemokine receptor expression among V alpha 24(+)V beta 11(+) NKT cell subsets with distinct cytokine-producing capacity.
Blood
Kim CH,Johnston B,Butcher EC
MA1-19129 was used in flow cytometry to report differential expression of chemokine receptors among NKT cell subsets.
Mon Jul 01 00:00:00 EDT 2002
Identity of Leu-19 (CD56) leukocyte differentiation antigen and neural cell adhesion molecule.
The Journal of experimental medicine
Lanier LL,Testi R,Bindl J,Phillips JH
MA1-19129 was used in immunoprecipitation to propose that the Leu-19 antigen on leukocytes contributes to cell adhesion and is analogous to N-CAM on neural cells.
Thu Jun 01 00:00:00 EDT 1989