Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CD58 (Met1-Arg215) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.
This gene encodes a member of the immunoglobulin superfamily. The encoded protein is a ligand of the T lymphocyte CD2 protein, and functions in adhesion and activation of T lymphocytes. The protein is localized to the plasma membrane. Alternatively spliced transcript variants have been described. [provided by RefSeq, Jan 2009]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ag3; CD58; CD58 antigen, (lymphocyte function-associated antigen 3); FLJ23181; FLJ43722; LAIR1; Lymphocyte function-associated antigen 3; RP5-1086K13.2; Surface glycoprotein LFA-3
Gene Aliases: ag3; CD58; LFA-3; LFA3
UniProt ID: (Human) P19256
Entrez Gene ID: (Human) 965
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support