Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody recognizes human CD65, an asialo-fucoganglioside expressed on the surface of peripheral blood granulocytes (highly) and monocytes (weakly).
This type 2 fucosylated carbohydrate antigen is expressed on the surface of all myeloid cells during development. Moreover, CD65 is expressed highly on granulocytes and weakly on monocytes in peripheral blood. CD65 has been reported to be a ligand for CD62E (E-Selectin). However, the sialylated form of CD65 (designated CD65s) can be used as a marker for acute myeloid leukemia (AML) as its expression increases as CD34 decreases. Finally, CD65 is important for extravascular infiltration of AML cells.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: Fucoganglioside Type 2; Fucoganglioside Type II