Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: This KOR-SA3544 monoclonal antibody reacts with human CD66c (also known as CEACAM6), which is one of the splice variants of CD66. CD66c is expressed on endothelial and epithelial cells, as well as on neutrophils and monocytes. Myeloma and cancer cells, such as B-cell acute lymphoblastic lymphomas, also express CD66c. Moreover, expression of this receptor can be induced on T cells, B cells, and CD16-negative NK cells. Like the other CD66 splice variants, CD66c is involved in cell adhesion and growth, neutrophil activation, and signaling. Moreover, expression of CD66c in leukemic cells has been correlated with BCR/ABL1 rearrangement. Finally, studies have shown that the CD66 variants interact with each other homo- and heterotypically.
Applications Reported: This KOR-SA3544 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This KOR-SA3544 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 µL (0.06 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
CEA Cell Adhesion Molecule 6 (CEACAM6) belongs to the carcinoembryonic antigen (CEA) family whose members are glycosyl phosphatidyl inositol (GPI) anchored cell surface glycoproteins. Members of this family play a role in cell adhesion and are widely used as tumor markers in serum immunoassay determinations of carcinoma. CEACAM6 affects the sensitivity of tumor cells to adenovirus infection. The acts as a receptor for adherent-invasive E. coli adhesion to the surface of ileal epithelial cells in patients with Crohn's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); CD66c; CEACAM-1; CEACAM-3; CEACAM-5; CEACAM-6; CGM1; Cluster of Differentiation 66c; Non-specific crossreacting antigen; Normal cross-reacting antigen; pan CEACAM
Gene Aliases: CD66c; CEACAM6; CEAL; NCA
UniProt ID: (Human) P40199
Entrez Gene ID: (Human) 4680
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support