Product References
SARS CoV-2 mRNA vaccination exposes latent HIV to Nef-specific CD8+ T-cells.
Nature communications
Stevenson EM,Terry S,Copertino D,Leyre L,Danesh A,Weiler J,Ward AR,Khadka P,McNeil E,Bernard K,Miller IG,Ellsworth GB,Johnston CD,Finkelsztein EJ,Zumbo P,Betel D,Dündar F,Duncan MC,Lapointe HR,Speckmaier S,Moran-Garcia N,Papa MP,Nicholes S,Stover CJ,Lynch RM,Caskey M,Gaebler C,Chun TW,Bosque A,Wilkin TJ,Lee GQ,Brumme ZL,Jones RB
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Fri Aug 19 00:00:00 EDT 2022
Cancer-secreted exosomal miR-1468-5p promotes tumor immune escape via the immunosuppressive reprogramming of lymphatic vessels.
Molecular therapy : the journal of the American Society of Gene Therapy
Zhou C,Wei W,Ma J,Yang Y,Liang L,Zhang Y,Wang Z,Chen X,Huang L,Wang W,Wu S
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Wed Apr 07 00:00:00 EDT 2021
Dysregulated Peripheral Invariant Natural Killer T Cells in Plaque Psoriasis Patients.
Frontiers in cell and developmental biology
Hu Y,Chen Y,Chen Z,Zhang X,Guo C,Yu Z,Xu P,Sun L,Zhou X,Gong Y,Yu Q,Shi Y
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Wed Feb 23 00:00:00 EST 2022
T cell receptor-dependent S-acylation of ZAP-70 controls activation of T cells.
The Journal of biological chemistry
Tewari R,Shayahati B,Fan Y,Akimzhanov AM
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Thu Aug 26 00:00:00 EDT 2021
Identification of celastrol as a novel HIV-1 latency reversal agent by an image-based screen.
PloS one
Liu H,Hu PW,Dubrulle J,Stossi F,Nikolai BC,Mancini MA,Rice AP
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Tue Sep 14 00:00:00 EDT 2021
Distinct populations of antigen-specific tissue-resident CD8+ T cells in human cervix mucosa.
JCI insight
Peng T,Phasouk K,Bossard E,Klock A,Jin L,Laing KJ,Johnston C,Williams NA,Czartoski JL,Varon D,Long AN,Bielas JH,Snyder TM,Robins H,Koelle DM,McElrath MJ,Wald A,Corey L,Zhu J
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Immunocytochemistry
Mon Aug 09 00:00:00 EDT 2021
Human endoglin-CD3 bispecific T cell engager antibody induces anti-tumor effect in vivo.
Theranostics
Zhong L,Shi W,Gan L,Liu X,Huo Y,Wu P,Zhang Z,Wu T,Peng H,Huang Y,Zhao Y,Yuan Y,Deng Z,Tang H
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Wed Jul 28 00:00:00 EDT 2021
Longitudinal single-cell profiling reveals molecular heterogeneity and tumor-immune evolution in refractory mantle cell lymphoma.
Nature communications
Zhang S,Jiang VC,Han G,Hao D,Lian J,Liu Y,Zhang R,McIntosh J,Wang R,Dang M,Dai E,Wang Y,Santos D,Badillo M,Leeming A,Chen Z,Hartig K,Bigcal J,Zhou J,Kanagal-Shamanna R,Ok CY,Lee H,Steiner RE,Zhang J,Song X,Nair R,Ahmed S,Rodriquez A,Thirumurthi S,Jain P,Wagner-Bartak N,Hill H,Nomie K,Flowers C,Futreal A,Wang L,Wang M
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Mon May 17 00:00:00 EDT 2021
Coagulation factor V is a marker of tumor-infiltrating immune cells in breast cancer.
Oncoimmunology
Tinholt M,Stavik B,Tekpli X,Garred Ø,Borgen E,Kristensen V,Sahlberg KK,Sandset PM,Iversen N
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Tue Sep 29 00:00:00 EDT 2020
Unique natural killer cell subpopulations are associated with exacerbation risk in chronic obstructive pulmonary disease.
Scientific reports
Osterburg AR,Lach L,Panos RJ,Borchers MT
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Mon Jan 27 00:00:00 EST 2020
Impact of food challenge on local oesophageal immunophenotype in eosinophilic oesophagitis.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology
Philpott H,Lee SZ,Arrington A,McGee SJ,Dellon ES
14-0699-82 was used in Immunohistochemistry to perform a comparative study of the density of cell surface proteins (known to be involved with antigen presentation) on oesophageal tissue prior to, and following food antigen induced disease recurrence.
Wed Apr 01 00:00:00 EDT 2020
Transcriptional down-regulation of ccr5 in a subset of HIV+ controllers and their family members.
eLife
Gonzalo-Gil E,Rapuano PB,Ikediobi U,Leibowitz R,Mehta S,Coskun AK,Porterfield JZ,Lampkin TD,Marconi VC,Rimland D,Walker BD,Deeks S,Sutton RE
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Tue Apr 09 00:00:00 EDT 2019
Heterogeneity of human bone marrow and blood natural killer cells defined by single-cell transcriptome.
Nature communications
Yang C,Siebert JR,Burns R,Gerbec ZJ,Bonacci B,Rymaszewski A,Rau M,Riese MJ,Rao S,Carlson KS,Routes JM,Verbsky JW,Thakar MS,Malarkannan S
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Mon Sep 02 00:00:00 EDT 2019
Intracellular delivery of mRNA to human primary T cells with microfluidic vortex shedding.
Scientific reports
Jarrell JA,Twite AA,Lau KHWJ,Kashani MN,Lievano AA,Acevedo J,Priest C,Nieva J,Gottlieb D,Pawell RS
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2019
NK-Cell Recruitment Is Necessary for Eradication of Peritoneal Carcinomatosis with an IL12-Expressing Maraba Virus Cellular Vaccine.
Cancer immunology research
Alkayyal AA,Tai LH,Kennedy MA,de Souza CT,Zhang J,Lefebvre C,Sahi S,Ananth AA,Mahmoud AB,Makrigiannis AP,Cron GO,Macdonald B,Marginean EC,Stojdl DF,Bell JC,Auer RC
14-0699-82 was used in Flow Cytometry to suggest that an MG1-IL12-ICV is a promising therapy that could provide benefit to the thousands of patients diagnosed with PC each year.
Wed Mar 01 00:00:00 EST 2017
Glossogyne tenuifolia (Hsiang-ju) extract suppresses T cell activation by inhibiting activation of c-Jun N-terminal kinase.
Chinese medicine
Houng JY,Tai TS,Hsu SC,Hsu HF,Hwang TS,Lin CJ,Fang LW
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
Hormonal vitamin D up-regulates tissue-specific PD-L1 and PD-L2 surface glycoprotein expression in humans but not mice.
The Journal of biological chemistry
Dimitrov V,Bouttier M,Boukhaled G,Salehi-Tabar R,Avramescu RG,Memari B,Hasaj B,Lukacs GL,Krawczyk CM,White JH
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Fri Dec 15 00:00:00 EST 2017
Combinations of isoform-targeted histone deacetylase inhibitors and bryostatin analogues display remarkable potency to activate latent HIV without global T-cell activation.
Scientific reports
Albert BJ,Niu A,Ramani R,Marshall GR,Wender PA,Williams RM,Ratner L,Barnes AB,Kyei GB
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Mon Aug 07 00:00:00 EDT 2017
MAIT cells are activated during human viral infections.
Nature communications
van Wilgenburg B,Scherwitzl I,Hutchinson EC,Leng T,Kurioka A,Kulicke C,de Lara C,Cole S,Vasanawathana S,Limpitikul W,Malasit P,Young D,Denney L,Moore MD,Fabris P,Giordani MT,Oo YH,Laidlaw SM,Dustin LB,Ho LP,Thompson FM,Ramamurthy N,Mongkolsapaya J,Willberg CB,Screaton GR,Klenerman P
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Thu Jun 23 00:00:00 EDT 2016
Drug conjugated nanoparticles activated by cancer cell specific mRNA.
Oncotarget
Gossai NP,Naumann JA,Li NS,Zamora EA,Gordon DJ,Piccirilli JA,Gordon PM
14-0699-82 was used in Flow Cytometry to develop a customisable approach to cancer therapy in which a gold nanoparticle delivers a drug that is selectively activated within the cancer cell by the presence of an mRNA unique to the cancer cell.
Tue Jun 21 00:00:00 EDT 2016
Functional dichotomy of Vδ2 γδ T cells in chronic hepatitis C virus infections: role in cytotoxicity but not for IFN-γ production.
Scientific reports
Yin W,Tong S,Zhang Q,Shao J,Liu Q,Peng H,Hu H,Peng M,Hu P,Ren H,Tian Z,Zhang D
14-0699 was used in Flow cytometry/Cell sorting to analyse the phenotype and function of Vδ2 T cells in HCV-infected patients compared to healthy controls.
Thu May 19 00:00:00 EDT 2016
Mucosal-associated invariant T cell alterations in obese and type 2 diabetic patients.
The Journal of clinical investigation
Magalhaes I,Pingris K,Poitou C,Bessoles S,Venteclef N,Kiaf B,Beaudoin L,Da Silva J,Allatif O,Rossjohn J,Kjer-Nielsen L,McCluskey J,Ledoux S,Genser L,Torcivia A,Soudais C,Lantz O,Boitard C,Aron-Wisnewsky J,Larger E,Clément K,Lehuen A
14-0699 was used in Flow cytometry/Cell sorting to analyse MAIT cells in the blood and adipose tissues of patients with type 2 diabetes and/or severe obesity.
Wed Apr 01 00:00:00 EDT 2015
HIV Tat controls RNA Polymerase II and the epigenetic landscape to transcriptionally reprogram target immune cells.
eLife
Reeder JE,Kwak YT,McNamara RP,Forst CV,D'Orso I
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Wed Oct 21 00:00:00 EDT 2015
RSV-specific airway resident memory CD8+ T cells and differential disease severity after experimental human infection.
Nature communications
Jozwik A,Habibi MS,Paras A,Zhu J,Guvenel A,Dhariwal J,Almond M,Wong EHC,Sykes A,Maybeno M,Del Rosario J,Trujillo-Torralbo MB,Mallia P,Sidney J,Peters B,Kon OM,Sette A,Johnston SL,Openshaw PJ,Chiu C
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Mon Dec 21 00:00:00 EST 2015
Higher Frequency of NK and CD4+ T-Cells in Mucosa and Potent Cytotoxic Response in HIV Controllers.
PloS one
Taborda NA,González SM,Alvarez CM,Correa LA,Montoya CJ,Rugeles MT
14-0699 was used in Flow cytometry/Cell sorting to investigate the frequency, phenotype and activity of T and NK cells in HIV controllers, showing higher frequencies of NK and CD4+ T-Cells and potent cytotoxic response.
Fri May 13 00:00:00 EDT 2016
Alternative capture of noncoding RNAs or protein-coding genes by herpesviruses to alter host T cell function.
Molecular cell
Guo YE,Riley KJ,Iwasaki A,Steitz JA
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Thu Apr 10 00:00:00 EDT 2014
Innate immune responses and rapid control of inflammation in African green monkeys treated or not with interferon-alpha during primary SIVagm infection.
PLoS pathogens
Jacquelin B,Petitjean G,Kunkel D,Liovat AS,Jochems SP,Rogers KA,Ploquin MJ,Madec Y,Barré-Sinoussi F,Dereuddre-Bosquet N,Lebon P,Le Grand R,Villinger F,Müller-Trutwin M
14-0699 was used in Flow cytometry/Cell sorting to investigate the role of innate immune cells and IFN-α in the control of chronic immune activation, showing that control is independent of IFN-α levels in SIV infection.
Tue Jul 01 00:00:00 EDT 2014
Apoptotic signaling through Fas and TNF receptors ameliorates GVHD in mobilized peripheral blood grafts.
Bone marrow transplantation
Mizrahi K,Yaniv I,Ash S,Stein J,Askenasy N
14-0699-82 was used in Flow Cytometry to assess the susceptibility of mobilised peripheral blood cells to spontaneous and receptor-induced apoptosis as a possible approach to GVHD prophylaxis.
Thu May 01 00:00:00 EDT 2014
Lipoxin A4 regulates natural killer cell and type 2 innate lymphoid cell activation in asthma.
Science translational medicine
Barnig C,Cernadas M,Dutile S,Liu X,Perrella MA,Kazani S,Wechsler ME,Israel E,Levy BD
14-0699 was used in Flow cytometry/Cell sorting to identify the potential of innate lymphoid cells to decrease airway inflammation.
Wed Feb 27 00:00:00 EST 2013
Ex vivo analysis of resident hepatic pro-inflammatory CD1d-reactive T cells and hepatocyte surface CD1d expression in hepatitis C.
Journal of viral hepatitis
Yanagisawa K,Yue S,van der Vliet HJ,Wang R,Alatrakchi N,Golden-Mason L,Schuppan D,Koziel MJ,Rosen HR,Exley MA
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Thu Aug 01 00:00:00 EDT 2013
Single cell microRNA analysis using microfluidic flow cytometry.
PloS one
Wu M,Piccini M,Koh CY,Lam KS,Singh AK
Published figure using CD69 monoclonal antibody (Product # 14-0699-82) in Flow Cytometry
Fri Jul 19 00:00:00 EDT 2013
Lung dendritic cell expression of maturation molecules increases with worsening chronic obstructive pulmonary disease.
American journal of respiratory and critical care medicine
Freeman CM,Martinez FJ,Han MK,Ames TM,Chensue SW,Todt JC,Arenberg DA,Meldrum CA,Getty C,McCloskey L,Curtis JL
14-0699 was used in Flow cytometry/Cell sorting to determine the expression of maturation molecules by individual DC subsets in relationship to COPD stage.
Tue Dec 15 00:00:00 EST 2009