Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 12
Immunogen sequence: QNVKHPVTG QFLYQDSNWA SKVEKLTLDN AAFPFLAYSG IPAVSFCFCE DTDYPYLGTT MDTYKELIER IPELNKVARA AAEVAGQFVI KLTHDVELNL DYERYNSQLL SFVRDLNQYR ADIKEMGLSL QWLYSARGDF FRATSRLTTD FGNAEKTDRF VMKKLNDRVM RVEYHFLSPY VSPKESPFRH VFWGSGSHTL PALLENLKLR KQNNGAFNET LFRNQLALAT WTIQGAANAL SG (511-751 aa encoded by BC001188)
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system. Serum transferrin receptor (sTfR) is used as a means of detecting erythropoietin (EPO) misuse by athletes and as a diagnostic test for anemias resulting from a number of conditions including rheumatoid arthritis, pregnancy, irritable bowel syndrome and in HIV patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD antigen CD71; CD71; p90; T9; TR; Transferrin receptor protein 1
Gene Aliases: CD71; IMD46; p90; T9; TFR; TFR1; TFRC; TR; TRFR
UniProt ID: (Human) P02786
Entrez Gene ID: (Human) 7037
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support