Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MGSACIKVTK YFLFLFNLIF FILGAVILGF GVWILADKSS FISVLQTSSS SLRMGAYVFI GVGAVTMLMG FLGCIGAVNE VRCLLGLYFA FLLLILIAQV TAGALFYFNM GKLKQEMGGI VTELIRDYNS SREDSLQDAW DYVQAQVKCC GWVSFYNWTD NAELMNRPEV TYPCSCEVKG EEDNSLSVRK GFCEAPGNRT QSGNHPEDWP VYQEGCMEKV QAWLQENLGI ILGVGVGVAI VELLGMVLSI CLCRHVHSED YSKVPKY
This metastasis suppressor gene product is a membrane glycoprotein that is a member of the transmembrane 4 superfamily. Expression of this gene has been shown to be downregulated in tumor progression of human cancers and can be activated by p53 through a consensus binding sequence in the promoter. Its expression and that of p53 are strongly correlated, and the loss of expression of these two proteins is associated with poor survival for prostate cancer patients. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C33 antigen; CD82; CD82 antigen; IA4; Inducible membrane protein R2; kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyte antigen, antigen detected by monoclonal and antibody IA4)); Metastasis suppressor Kangai-1; Suppressor of tumorigenicity 6 protein; Tetraspanin-27; Tspan-27
Gene Aliases: 4F9; C33; CD82; GR15; IA4; KAI1; R2; SAR2; ST6; TSPAN27
UniProt ID: (Human) P27701
Entrez Gene ID: (Human) 3732
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support