Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: The A2MR-a2 monoclonal antibody reacts with human CD91, also known as LRP-1 and alpha-2 macroglobulin receptor. CD91 is a 600 kDa, type 1 transmembrane protein consisting of an alpha and beta chain. The alpha chain interacts with multiple structurally and functionally diverse ligands and the beta chain contains 2 tyrosine phosphorylation motifs capable of interacting with adaptor and signaling proteins. Binding of ligands mediates ligand endocytosis and trafficking to lysosomes. This receptor is expressed by cells in the monocytic lineage, dendritic cells, hepatocytes, firbroblasts, adipocytes, neurons, and syncytiotrophoblast. CD91 is involved in cross-presenting antigens via MHC class I molecules on antigen presenting cells after endocytosis of heat shock protein-peptide complexes. It also plays a role in cell migration and is involved in activating signaling cascades.
Each product contains 1 vial of NovaFluor conjugate and 1 vial of CellBlox Plus Blocking Buffer .
Applications Reported: This A2MR-a2 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This A2MR-a2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.05 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
NovaFluor dyes are not compatible with DNA intercalating viability dyes. Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes.
This NovaFluor conjugate has been updated to ship with CellBlox Plus Blocking Buffer (Cat. No. (C001T06F01)). This buffer contains formulation improvements over CellBlox. CellBlox Plus Blocking Buffer is required for optimal staining with NovaFluor conjugates and should be used in all experiments where NovaFluor conjugates are used. Whenever possible, we recommend adding CellBlox Plus Blocking Buffer to antibody cocktails/master mixes prior to combining with cells. Add 5 µL per sample (regardless of the number of NovaFluors in your panel) to use the antibody cocktail as intended. For single-color controls, use 5 µL of CellBlox Blocking Buffer per 100 µL of cell sample containing 10^3 to 10^8 cells.
NovaFluor conjugates are based on Phiton™ technology utilizing novel nucleic acid dye structures that allow for engineered fluorescent signatures with consideration for spillover and spread impacts. Learn more
Excitation: 636 nm; Emission: 727 nm; Laser: 633-640 nm (Red) Laser
The protein encoded by this gene is an endocytic receptor involved in several cellular processes, including intracellular signaling, lipid homeostasis, and clearance of apoptotic cells. In addition, the encoded protein is necessary for the A2M-mediated clearance of secreted amyloid precursor protein and beta-amyloid, the main component of amyloid plaques found in Alzheimer patients. Expression of this gene decreases with age and has been found to be lower than controls in brain tissue from Alzheimer patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A2MR; Alpha-2-macroglobulin receptor; APOER; Apolipoprotein E receptor; CD91; FLJ16451; low density lipoprotein receptor-related protein 1; LRP-1; MGC88725; Prolow-density lipoprotein receptor-related protein 1; TbetaR-V/LRP-1/IGFBP-3 receptor; type V tgf-beta receptor
Gene Aliases: A2MR; APOER; APR; CD91; IGFBP3R; LRP; LRP1; LRP1A; TGFBR5
UniProt ID: (Human) Q07954
Entrez Gene ID: (Human) 4035
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support